Skip to Content
Merck
All Photos(2)

Documents

C104809

Sigma-Aldrich

Phenylcyclohexane

≥97%

Synonym(s):

Cyclohexylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H11
CAS Number:
Molecular Weight:
160.26
Beilstein:
1906803
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

liquid

refractive index

n20/D 1.526 (lit.)

bp

239-240 °C (lit.)

mp

4-7 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

C1CCC(CC1)c2ccccc2

InChI

1S/C12H16/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1,3-4,7-8,12H,2,5-6,9-10H2

InChI key

IGARGHRYKHJQSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L S Welch et al.
Toxicology and industrial health, 8(4), 47-50 (1992-07-01)
Investigation of this outbreak raises some important points for future research. Although for various reasons the case ascertainment for MCS was not complete, the three MCS patients described here all had preexisting conditions that may have put them at risk.
M P Holsapple et al.
Life sciences, 31(8), 803-813 (1982-08-23)
Initial acute behavioral studies in mice indicated that phencyclidine (PCP) produced marked motor impairment as measured by the inverted screen technique with an ED50 value of 4.1 muMole/kg (i.v.). Phenylcyclohexene (PC) was considerably less active with an ED50 value of
Eiichi Hayashi et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 52(2), 112-116 (2011-04-26)
In order to verify the safety associated with reusing PET and glass bottles, a challenge test was conducted with five surrogate contaminants: 1,1,1-trichloroethane, chlorobenzene, toluene, benzophenone and phenyl cyclohexane. Bottles were filled with a cocktail solution of these contaminants and
Athanasios Katsoyiannis et al.
Journal of hazardous materials, 152(2), 669-676 (2007-09-15)
Emissions of Volatile Organic Compounds (VOCs) and carbonyls from carpets of different type (wool, synthetic) over a time period of three days at 23 degrees C, at 45% relative humidity, 0.5 air change rate and a loading factor of 0.4
Brian D Fitz et al.
Journal of chromatography. A, 1392, 82-90 (2015-03-31)
Low thermal mass gas chromatography (LTM-GC) was evaluated for rapid, high peak capacity separations with three injection methods: liquid, headspace solid phase micro-extraction (HS-SPME), and direct vapor. An Agilent LTM equipped with a short microbore capillary column was operated at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service