Skip to Content
Merck
All Photos(4)

Documents

B103209

Sigma-Aldrich

2-Butyne-1,4-diol

99%

Synonym(s):

1,4-Butynediol, 1,4-Dihydroxy-2-butyne, 2-Butynediol, Bis(hydroxymethyl)acetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C≡CCH2OH
CAS Number:
Molecular Weight:
86.09
Beilstein:
1071237
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<0.1 mmHg ( 55 °C)

Quality Level

Assay

99%

form

crystals

bp

238 °C (lit.)

mp

53-58 °C (lit.)

SMILES string

OCC#CCO

InChI

1S/C4H6O2/c5-3-1-2-4-6/h5-6H,3-4H2

InChI key

DLDJFQGPPSQZKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2-Butyne-1,4-diol is extensively used in the cycloaddition reactions such as the homologation method for the preparation of substituted acenes, rhodium and iridium-catalyzed [2+ 2+ 2] inter and intramolecular cyclotrimerization. It can also be used in the total synthesis of (−)-isolaurallene, (−)-amphidinolide P and bistramide A.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

305.6 °F - closed cup

Flash Point(C)

152 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homologation method for preparation of substituted pentacenes and naphthacenes.
Takahashi T, et al.
The Journal of Organic Chemistry, 71(21), 7967-7977 (2006)
Total synthesis of (?)-amphidinolide P.
Williams D R, et al.
Organic Letters, 2(7), 945-948 (2000)
R A Jedrychowski et al.
Journal of applied toxicology : JAT, 12(2), 117-122 (1992-04-01)
2-Butyne-1,4-diol was given to male and female Wistar Imp:DAK rats by oral gavage for 28 consecutive days in daily doses of 1, 10 or 50 mg kg-1 day-1. After 28 days all animals were necropsied. Blood samples were obtained and
Małgorzata Kupczewska-Dobecka et al.
Medycyna pracy, 60(5), 347-357 (2009-12-17)
Derived No Effect Level (DNEL(inh)) has been set for occupational exposure to but-2-yno-1,4-diol according to REACH principles. Maximum allowable concentration (MAC) and DNEL(inh) have been compared. Experimental data from two inhalation studies on rats and three oral studies have been
J Hellwig et al.
Toxicology letters, 92(3), 221-230 (1997-08-22)
Developmental toxicity of 2-butin-1,4-diol was determined in groups of 18-22 pregnant Wistar rats at dose levels of 10, 40 and 80 mg/kg bw/day administered by gavage from days 6 to 15 pc. At 80 mg/kg bw/day food consumption and maternal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service