Skip to Content
Merck
All Photos(3)

Key Documents

A74807

Sigma-Aldrich

4-Aminophenyl sulfone

97%

Synonym(s):

4,4′-Diaminodiphenyl sulfone, 4,4′-Sulfonyldianiline, Bis(4-aminophenyl) sulfone, DDS, Dapsone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2NC6H4)2SO2
CAS Number:
Molecular Weight:
248.30
Beilstein:
788055
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

crystals

mp

175-177 °C (lit.)

SMILES string

Nc1ccc(cc1)S(=O)(=O)c2ccc(N)cc2

InChI

1S/C12H12N2O2S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8H,13-14H2

InChI key

MQJKPEGWNLWLTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-aminophenyl sulfone (DDS) is a diamine-based hardener that is used to cure epoxy-based resins.

Application

Tetrafunctional epoxy resins can be blended with DDS and can be thermally cured which can be used in oil field applications. DDS can also be used to cure iron oxide (Fe2O3) layer double hydroxide based epoxy that forms a flame retardant epoxy nanocomposites. It can also be used as a hardener to cure bisphenol A (DGEBA) for use in the fabrication of flame retardant epoxy/clay nanocomposites.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 1B - STOT RE 2 - STOT SE 2

Target Organs

Blood, Blood,spleen,Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glass transition dependence of ultrahigh strain rate response in amine cured epoxy resins.
Knorr Jr DB.
Polymer, 53(25), 5917-5923 (2012)
A novel high performance bismaleimide/diallyl bisphenol A (BMI/DBA)-epoxy interpenetrating network resin for rigid riser application.
Jena RK, et al.
Royal Society of Chemistry Advances, 5(97), 79888-79897 (2015)
Jaesung Seo et al.
Health and quality of life outcomes, 13, 138-138 (2015-09-05)
This paper aimed to employ subject matter experts (SMEs) to assess the extent to which the Korean version of the short-form of the OHIP (OHIP-14 K) is culturally valid and equivalent in Korean. We approached 17 bilingual Korean SMEs from
Ernest Güell et al.
Palliative & supportive care, 13(2), 295-303 (2014-04-26)
We aimed to address the prevalence of desire-to-die statements (DDSs) among terminally ill cancer patients in an acute palliative care unit. We also intended to compare the underlying differences between those patients who make desire-to-die comments (DDCs) and those who
F-R Zhang et al.
The New England journal of medicine, 369(17), 1620-1628 (2013-10-25)
Dapsone is used in the treatment of infections and inflammatory diseases. The dapsone hypersensitivity syndrome, which is associated with a reported mortality of 9.9%, develops in about 0.5 to 3.6% of persons treated with the drug. Currently, no tests are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service