Skip to Content
Merck
All Photos(1)

Key Documents

92751

Sigma-Aldrich

1-(Trimethylsilyl)imidazole

≥98.0%

Synonym(s):

N-Trimethylsilylimidazole, TMSI, TSIM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2Si
CAS Number:
Molecular Weight:
140.26
Beilstein:
606148
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

synthesis grade

Assay

≥98.0% (T)
≥98.0%

form

liquid

refractive index

n20/D 1.475 (lit.)

bp

93-94 °C/14 mmHg (lit.)

density

0.957 g/mL at 20 °C
0.956 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)n1ccnc1

InChI

1S/C6H12N2Si/c1-9(2,3)8-5-4-7-6-8/h4-6H,1-3H3

InChI key

YKFRUJSEPGHZFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-(Trimethylsilyl)imidazole may be used as an effective alternative to N,O-bis(trimethylsilyl)trifluoroacetamide (BSTFA) and N-methyl-N-(trimethylsilyl)trifluoroacetamide (MSTFA) for the efficient silylation of Ti-MCM-41, a titanium substituted mesoporous molecular sieve. This process enhances the hydrophobicity and catalytic activity of Ti-MCM-41 catalyst for cyclohexene epoxidation with aqueous hydrogen peroxide.

Other Notes

Powerful silylating agent, particularly for alcohols; Synthesis of acyl imidazolides

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lukas Schmidt et al.
Archives of toxicology, 91(2), 677-687 (2015-12-19)
We studied the human in vivo metabolism and the elimination kinetics of α-pinene (αPN), a natural monoterpene which commonly occurs in the environment. Four volunteers were exposed to a single oral dose of 10 mg αPN. Each subject provided one pre-exposure
Silylation of Ti?MCM?41 by trimethylsilyl?imidazole and its effect on the olefin epoxidation with aqueous H 2 O 2.
Bu J & Rhee HK
Catalysis Letters, 66(4), 245-249 (2000)
J. Heberle et al.
Silylating Agents, 2nd ed. (1995)
J.D. Nicholson
Analyst, 103, 193-193 (1978)
G.S. Bates et al.
Tetrahedron Letters, 4423-4423 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service