Skip to Content
Merck
All Photos(1)

Key Documents

900374

Sigma-Aldrich

α-Tocopherol-(phenyl-13C6)

≥99 atom % 13C, ≥96% (CP)

Synonym(s):

Vitamin E-(phenyl-13C6)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C23H50O2
Molecular Weight:
436.66
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % 13C

Assay

≥96% (CP)

form

liquid

technique(s)

mass spectrometry (MS): suitable

color

yellow to brown

mass shift

M+3
M+6

storage temp.

−20°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCCC1(C)CC[13C]2=[13C](C)[13C](O)=[13C](C)[13C](C)=[13C]2O1

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/t21-,22-,29?/m1/s1/i23+1,24+1,25+1,26+1,27+1,28+1

InChI key

GVJHHUAWPYXKBD-CVPWOIPRSA-N

General description

α-Tocopherol (vitamin E) is a lipid-soluble antioxidant found in whole grains, nuts, and oils. It is known to scavenge reactive oxygen species by interacting with lipids during oxidative stress.
α-Tocopherol-(phenyl-13C6) is an isotope of vitamin E wherein all the six phenyl carbons are replaced by 13C6 carbon.

Application

α-Tocopherol-(phenyl-13C6) can be used as a stable isotope internal standard for data interpretation in specific biological samples.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ANTIOXIDANTS-Role of Antioxidant Nutrients in Defense Systems
Encyclopedia of food sciences and nutrition, 282-289 (2003)
The role of alpha-tocopherol in plant stress tolerance
Munne-Bosch S
Journal of Plant Physiology, 162(7), 743-748 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service