Skip to Content
Merck
All Photos(1)

Key Documents

900369

Sigma-Aldrich

(±)-Epigallocatechin-2,3,4-13C3

≥99 atom % 13C, ≥97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C12H14O7
Molecular Weight:
309.25
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % 13C

Assay

≥97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

SMILES string

OC(C=C1O)=CC2=C1[13C][13C](O)[13C](C3=CC(O)=C(O)C(O)=C3)O2

InChI

1S/C15H10O7/c16-7-3-9(17)8-5-12(20)15(22-13(8)4-7)6-1-10(18)14(21)11(19)2-6/h1-4,16-21H/i5+1,12+1,15+1

InChI key

CKSTYZPHICYKHH-HQTPQMSASA-N

Related Categories

Application

Relative stereochemistry

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service