Skip to Content
Merck
All Photos(1)

Key Documents

809322

Sigma-Aldrich

1-(2-Aminoethyl)maleimide hydrochloride

Synonym(s):

1-(2-Aminoethyl)pyrrole-2,5-dione hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N2O2Cl
Molecular Weight:
176.60
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

Quality Level

form

powder

mp

166 °C

functional group

amine
imide
maleimide

SMILES string

Cl.N1(C(=O)C=CC1=O)CCN

InChI

1S/C6H8N2O2.ClH/c7-3-4-8-5(9)1-2-6(8)10;/h1-2H,3-4,7H2;1H

InChI key

NJQOCRDPGFWEKA-UHFFFAOYSA-N

General description

1-(2-Aminoethyl)maleimide hydrochloride (N-(2-Aminoethyl)maleimide hydrochloride salt) can be prepared from tert-butyl 2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethylcarbamate.[1]

Application

1-(2-Aminoethyl)maleimide hydrochloride (1-(2-Aminoethyl)-1H-pyrrole-2,5-dione HCl salt) may be used in the preparation of N-propionyl cysteaminylphenol-maleimide-dextran (NPCMD)[2] and 4-nitrophenyl 2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethylcarbamate.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pieter van der Veken et al.
Chembiochem : a European journal of chemical biology, 6(12), 2271-2280 (2005-10-29)
Gaining insight into phosphoproteomes is of the utmost importance for understanding regulation processes such as signal transduction and cellular differentiation. While the identification of phosphotyrosine-containing amino acid sequences in peptides and proteins is now becoming possible, mainly because of the
Yu Mizote et al.
Journal of dermatological science, 73(3), 209-215 (2013-12-10)
N-propionyl cysteaminylphenol-maleimide-dextran (NPCMD) is a toxic tyrosinase substrate developed to treat melanoma. We investigated the effect of NPCMD on innate immune responses in monocytes. CD14⁺ monocytes and a monocytic cell line, THP-1, were stimulated with NPCMD in vitro. Cytokines in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service