Skip to Content
Merck
All Photos(2)

Key Documents

727504

Sigma-Aldrich

4-Amino-2-iodophenol

≥95.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6INO
CAS Number:
Molecular Weight:
235.02
Beilstein:
2716290
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (HPLC)

form

powder

composition

carbon, 29.1-32.2% elemental analysis
nitrogen, 5.7-6.3% elemental analysis

mp

110-120 °C (subl.) (lit.)

SMILES string

Nc1ccc(O)c(I)c1

InChI

1S/C6H6INO/c7-5-3-4(8)1-2-6(5)9/h1-3,9H,8H2

InChI key

GUGSDWSSHLBVST-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service