Skip to Content
Merck
All Photos(1)

Key Documents

700673

Sigma-Aldrich

(R)-3,3′-Di-9-phenanthrenyl-1,1′-bi-2-naphthol

97%

Synonym(s):

(R)-9-Phenanthryl BINOL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H30O2
Molecular Weight:
638.75
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

optical activity

[α]22/D +62°, c = 1 in chloroform

mp

192-202 °C

storage temp.

−20°C

SMILES string

Oc1c(cc2ccccc2c1-c3c(O)c(cc4ccccc34)-c5cc6ccccc6c7ccccc57)-c8cc9ccccc9c%10ccccc8%10

InChI

1S/C48H30O2/c49-47-43(41-25-29-13-1-5-17-33(29)37-21-9-11-23-39(37)41)27-31-15-3-7-19-35(31)45(47)46-36-20-8-4-16-32(36)28-44(48(46)50)42-26-30-14-2-6-18-34(30)38-22-10-12-24-40(38)42/h1-28,49-50H

InChI key

XYUOTCUHTHQCNC-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service