Skip to Content
Merck
All Photos(2)

Key Documents

695890

Sigma-Aldrich

Phosphoric acid 2-hydroxyethyl methacrylate ester

contains 700-1000 ppm monomethyl ether hydroquinone, 90%

Synonym(s):

2-Hydroxyethyl methacrylate phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10O3 · xH3PO4
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

~25% diester content

Quality Level

Assay

90%

form

liquid

contains

700-1000 ppm monomethyl ether hydroquinone

refractive index

n20/D 1.4688

density

1.37 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[P](=O)(O)(O)O.O(CCO)C(=O)C(=C)C

InChI

1S/C6H10O3.H3O4P/c1-5(2)6(8)9-4-3-7;1-5(2,3)4/h7H,1,3-4H2,2H3;(H3,1,2,3,4)

InChI key

POLZHVHESHDZRD-UHFFFAOYSA-N

General description

Phosphoric acid 2-hydroxyethyl methacrylate ester is a phosphoric acid based ester which is composed of phosphoric acid monomer and diethyl methacrylate. It can be used as a chelating absorbent due to its high affinity towards metal ions.

Application

Phosphoric acid 2-hydroxyethyl methacrylate ester is used in surface functionalization of polytetrafluoroethylene (PTFE) for craniofacial applications.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

293.0 °F - open cup

Flash Point(C)

145 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid removal of arsenic (V) by zirconium (IV) loaded phosphoric chelate adsorbent synthesized by radiation induced graft polymerization
Seko N, et al.
Reactive and Functional Polymers, 59(3), 235-241 (2004)
Deposition of a hydroxyapatite thin layer onto a polymer surface carrying grafted phosphate polymer chains.
K Kato et al.
Journal of biomedical materials research, 32(4), 687-691 (1996-12-01)
Y Tamada et al.
Journal of biomaterials science. Polymer edition, 10(7), 787-793 (1999-07-30)
Silk fabric was modified with polymethacryloyloxyethylphosphate (pMOEP) by graft copolymerization. Ca-adsorption onto pMOEP-grafted silk fabric was significantly enhanced compared to that onto original silk fabric. SEM observation indicated that some crystallites were deposited on the pMOEP-grafted silk fabric after 1
Influence of self-etching primer treatment on the adhesion of resin composite to polished dentin and enamel
Hayakawa T, et al.
Dental Materials : Official Publication of the Academy of Dental Materials, 14(2), 99-105 (1998)
K de Groot et al.
Journal of dental research, 59(9), 1493-1496 (1980-09-01)
The mechanical properties of two commercially available dental cements, a zinc phosphate cement and a carboxylate cement, were substantially improved by adding a polymeric phosphate acid to the cement liquids. The improvements can be explained in terms of the change

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service