Skip to Content
Merck
All Photos(1)

Documents

674311

Sigma-Aldrich

11-Mercaptoundecylphosphoric acid

90%

Synonym(s):

11-Mercapto-1-undecyl dihydrogen phosphate, MDPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H25O4PS
CAS Number:
Molecular Weight:
284.35
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

90%

form

solid

mp

48-52 °C

storage temp.

2-8°C

SMILES string

OP(O)(=O)OCCCCCCCCCCCS

InChI

1S/C11H25O4PS/c12-16(13,14)15-10-8-6-4-2-1-3-5-7-9-11-17/h17H,1-11H2,(H2,12,13,14)

InChI key

XPYXJSSCWCSOFO-UHFFFAOYSA-N

Application

High purity phosphate derivitized thiol for SAMs

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine A McIntyre et al.
Talanta, 210, 120188-120188 (2020-01-29)
Myo-inositol hexakisphosphate, or phytic acid, (myo-IP6) is a key organic phosphorus (P) compound in soils and manures. Determinations of myo-IP6 in soils and manure extracts are frequently performed by 31P NMR spectroscopy. This approach is time-consuming in terms of both
Ann E Fink et al.
PloS one, 7(8), e41434-e41434 (2012-08-08)
Minimally invasive measurements of neuronal activity are essential for understanding how signal processing is performed by neuronal networks. While optical strategies for making such measurements hold great promise, optical sensors generally lack the speed and sensitivity necessary to record neuronal
Meital Eckshtain-Levi et al.
Nature communications, 7, 10744-10744 (2016-02-27)
Chirality-induced spin selectivity is a recently-discovered effect, which results in spin selectivity for electrons transmitted through chiral peptide monolayers. Here, we use this spin selectivity to probe the organization of self-assembled α-helix peptide monolayers and examine the relation between structural
Tatsuro Goda et al.
Micromachines, 11(9) (2020-09-03)
Chemical and enzymatic modifications of amino acid residues in protein after translation contain rich information about physiological conditions and diseases. Histone acetylation/deacetylation is the essential post-translational modification by regulating gene transcription. Such qualitative changes of biomacromolecules need to be detected
Alissa H Brandes et al.
Breast cancer research : BCR, 12(5), R84-R84 (2010-10-16)
17-allyamino-17-demethoxygeldanamycin (17-AAG), a small molecule inhibitor of Hsp90, is currently in clinical trials in breast cancer. However, 17-AAG treatment often results in inhibition of tumor growth rather than shrinkage, making detection of response a challenge. Magnetic resonance spectroscopy (MRS) and

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service