Skip to Content
Merck
All Photos(4)

Key Documents

633348

Sigma-Aldrich

Vinylboronic acid pinacol ester

contains phenothiazine as stabilizer, 95%

Synonym(s):

2-Ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaoborolane, 4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15BO2
CAS Number:
Molecular Weight:
154.01
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

contains

phenothiazine as stabilizer

refractive index

n20/D 1.4300 (lit.)

density

0.908 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CC1(C)OB(OC1(C)C)C=C

InChI

1S/C8H15BO2/c1-6-9-10-7(2,3)8(4,5)11-9/h6H,1H2,2-5H3

InChI key

DPGSPRJLAZGUBQ-UHFFFAOYSA-N

Application

Employed in a "double" Heck-Mizoroki arylation leading to β,β-diarylated vinyl boronates which react with an additional aryl halide to form Π-extended systems. This approach was used to prepare conjugated dendrimers. Also used to prepare γ-carbonyl vinyl boronates via a light-induced radical addition of xanthates.
Reagent used for
  • Suzuki-Miyaura coupling reactions
  • Mizoroki-Heck reactions (cascade reaction)
  • Intramolecular Nozaki-Hiyama-Kishi reactions
  • Stereoselective Cu-catalyzed γ-selective and stereospecific coupling
  • Control of stereoselectivity and mechanistic portrait on intramolecular (4+1)-cycloaddition of dialkoxycarbenes
  • Regio- and stereoselective synthesis of trisubstituted alkenes via gold(I)-catalyzed hydrophosphoryloxylation of haloalkynes followed by Pd-catalyzed consecutive cross-coupling reactions
  • Asymmetric Birch reductive alkylation

Reagent used in Preparation of
  • Molecular tubes for lipid sensing
  • Enzymatic inhibitors, antibiotics, receptor analogs, and other biologically significant compounds (including total syntheses)

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F

Flash Point(C)

34 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christopher A Leclair et al.
Tetrahedron letters, 51(52), 6852-6855 (2011-04-26)
A total synthesis of LL-Z1640-2 (2), a potent and selective kinase inhibitor, has been completed. The key step of the convergent synthesis utilized a late-stage intramolecular Nozaki-Hiyama-Kishi (NHK) reaction to close the macrocycle at the C6'-C7' bond.
Bathoju Chandra Chary et al.
Chemical communications (Cambridge, England), 47(27), 7851-7853 (2011-06-07)
A new stereoselective synthesis of trisubstituted alkenes is developed. Hydrophosphoryloxylation of haloalkynes provides Z-alkenyl halophosphates, which undergo Pd-catalyzed consecutive cross-coupling reactions to afford regio- and stereodefined trisubstituted alkenes.
Markus R Heinrich et al.
Chemical communications (Cambridge, England), (24), 3077-3079 (2005-06-17)
Gamma-carbonyl vinyl boronates can be prepared by a visible light induced radical chain addition of an S-acyl dithiocarbonate (xanthate) to the pinacol ester of vinyl boronic acid, followed by treatment with base.
Leslie Duroure et al.
Organic & biomolecular chemistry, 9(23), 8112-8118 (2011-10-26)
Simple models of the spiroimine core of (-)-gymnodimine A have been synthesized in racemic and optically active forms. The quaternary carbon of the racemic spiroimines was created by Michael addition of a β-ketoester to acrolein, whereas the asymmetric allylic alkylation
Christopher T Avetta et al.
The Journal of organic chemistry, 77(2), 851-857 (2012-01-24)
Two fluorescent sensors for lipids have been prepared and tested for detection of a number of hydrophobic compounds of varying shape and size. The data suggest that the two sensors have a different mode of fluorescent response. Yet, the two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service