616796
Salicylic acid-d6
98 atom % D, 99% (CP)
Synonym(s):
2-Hydroxybenzoic acid-d6
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
isotopic purity
98 atom % D
Assay
99% (CP)
bp
211 °C
mp
158-161 °C
mass shift
M+6
SMILES string
[2H]OC(=O)c1c([2H])c([2H])c([2H])c([2H])c1O[2H]
InChI
1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)/i1D,2D,3D,4D/hD2
InChI key
YGSDEFSMJLZEOE-UDDMDDBKSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
314.6 °F - closed cup
Flash Point(C)
157 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Michael A Falk et al.
Oecologia, 187(2), 471-482 (2018-05-26)
Empirical studies indicate that host-tree bud break will likely advance faster than spring-folivore egg hatch in response to predicted increases in temperature. How these phenological shifts will affect herbivory will depend on temporal patterns of foliar traits that occur during
Benny Björkblom et al.
British journal of cancer, 122(2), 221-232 (2019-12-11)
High-grade gliomas are associated with poor prognosis. Tumour heterogeneity and invasiveness create challenges for effective treatment and use of systemically administrated drugs. Furthermore, lack of functional predictive response-assays based on drug efficacy complicates evaluation of early treatment responses. We used
Kristen Van Gelder et al.
Planta, 252(1), 3-3 (2020-06-10)
A stable isotope-assisted mass spectrometry-based platform was utilized to demonstrate that the plant hormone, salicylic acid, is catabolized to catechol, a widespread secondary plant compound. The phytohormone salicylic acid (SA) plays a central role in the overall plant defense program
Junsik Choi et al.
Plant physiology, 179(4), 1315-1329 (2019-01-31)
Defects in the nuclear lamina of animal cell nuclei have dramatic effects on nuclear structure and gene expression as well as diverse physiological manifestations. We report that deficiencies in CROWDED NUCLEI (CRWN), which are candidate nuclear lamina proteins in Arabidopsis
Stacy A Voils et al.
Thrombosis research, 165, 6-13 (2018-03-16)
Incidence of venous thromboembolism (VTE) in critically ill patients remains unacceptably high despite widespread use of thromboprophylaxis. A systems biology approach may be useful in understanding disease pathology and predicting response to treatment. Metabolite profile under specific environmental conditions provides
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service