Skip to Content
Merck
All Photos(1)

Key Documents

605190

Sigma-Aldrich

DL-Homocystine-1,1′-13C2

99 atom % 13C

Synonym(s):

DL-Homocysteine dimer-1,1′-13C2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C6N2S2O4H16
Molecular Weight:
270.34
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

solid

mass shift

M+2

SMILES string

NC(CCSSCCC(N)[13C](O)=O)[13C](O)=O

InChI

1S/C8H16N2O4S2/c9-5(7(11)12)1-3-15-16-4-2-6(10)8(13)14/h5-6H,1-4,9-10H2,(H,11,12)(H,13,14)/i7+1,8+1

InChI key

ZTVZLYBCZNMWCF-BFGUONQLSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Rea et al.
The Journal of biological chemistry, 273(30), 18784-18792 (1998-07-21)
In adipocytes, insulin stimulates the translocation of the glucose transporter, GLUT4, from an intracellular storage compartment to the cell surface. Substantial evidence exists to suggest that in the basal state GLUT4 resides in discrete storage vesicles. A direct interaction of
Helen R McPherson et al.
eLife, 10 (2021-10-12)
Fibrinogen is essential for blood coagulation. The C-terminus of the fibrinogen α-chain (αC-region) is composed of an αC-domain and αC-connector. Two recombinant fibrinogen variants (α390 and α220) were produced to investigate the role of subregions in modulating clot stability and
Kevin Tröndle et al.
Journal of tissue engineering and regenerative medicine, 13(10), 1883-1895 (2019-07-18)
Active nutrient supply and waste product removal are key requirements for the fabrication of long-term viable and functional tissue constructs of considerable size. This work aims to contribute to the fabrication of artificial perfusable networks with a bioprinting process, based
S M Van Patten et al.
The Journal of biological chemistry, 274(15), 10268-10276 (1999-04-03)
Commercially available human plasma-derived preparations of the serine protease inhibitor antithrombin (AT) were shown to contain low levels of oxidation, and we sought to determine whether oxidation might be a means of regulating the protein's inhibitory activity. A recombinant form
Vijaykumar S Meli et al.
Science advances, 6(49) (2020-12-06)
Macrophages are innate immune cells that adhere to the extracellular matrix within tissues. However, how matrix properties regulate their function remains poorly understood. Here, we report that the adhesive microenvironment tunes the macrophage inflammatory response through the transcriptional coactivator YAP.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service