Skip to Content
Merck
All Photos(1)

Documents

576832

Sigma-Aldrich

Silver

nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis

Synonym(s):

Colloidal nano silver, Colloidal silver, Nanosilver

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
EC Number:
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23

Assay

99.5% trace metals basis

form

nanopowder

contains

PVP as dispersant

resistivity

1.59 μΩ-cm, 20°C

surface area

5.0 m2/g

particle size

<100 nm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

General description

This silver nanopowder consists of metallic silver nanoparticles with a diameter less than 100 nm. These nanosilver particles are coated with polyvinylpyrrolidone (PVP) at a concentration of 0.2% by mass. This coating effectively limits surface oxidation and enhances the dispersion of the powder in various solvents.

Silver nanopowder is extensively utilized in medicinal applications and medical devices. The increased surface area to volume ratio and heightened reactivity of nanosilver make it particularly valuable in modern biomedical applications, particularly in coatings of medical devices.

Application

Water based ink was formulated with the silver nanowires. Silver nanoparticles, leading to higher amounts of elutable substances has been studied. The immunological adjuvant effect of silver nanoparticles (AgNPs) was investigated both in vitro and in vivo Silver nanoparticles based graphite epoxy nanocomposites have been fabricated. Action of AgNPs in enhanced thrombus formation as a consequence of increased platelet aggregation and procoagulant activity has been investigated. Optical fiber sensor photodeposited with AgNPs have been fabricated. PAN fiber based nanocomposites containing Ag and Hydroxyapatite as nanoadditives have been reported.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silver nanocomposite electrode modified with hexacyanoferrate. Preparation, characterization and electrochemical behaviour towards substituted anilines
Fenga PG, et al.
Electroanalysis, 23(5), 1100-1106 (2011)
Silver nanoparticles toxicity and bactericidal effect against methicillin-resistant Staphylococcus aureus: nanoscale does matter
Ayala-Nu?ez et al.
NanoBiotechnology, 5(1-4), 2-9 (2009)
Yingying Xu et al.
Toxicology letters, 219(1), 42-48 (2013-03-05)
The immunological adjuvant effect of silver nanoparticles (AgNPs) was investigated both in vitro and in vivo. The in vivo adjuvant effect of AgNPs was evaluated with model antigen ovalbumin (OVA) and bovine serum albumin (BSA) in mice by intraperitoneal and
Antibacterial Properties of Silver-Finished Textiles
Matyjas-Zgondek E, et al.
Fibres and Textiles in Eastern Europe, 16(5), 101-107 (2008)
Optical Fiber Sensor Based on Localized Surface Plasmon Resonance Using Silver Nanoparticles Photodeposited on the Optical Fiber End
Ortega-Mendoza JG, et al.
Sensors, 14(10), 18701-18710 (2014)

Articles

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service