Skip to Content
Merck
All Photos(1)

Documents

54913

Sigma-Aldrich

Lithium β-hydroxypyruvate hydrate

≥97.0% (calc. based on dry substance, NT)

Synonym(s):

β-Hydroxypyruvic acid lithium salt hydrate, 3-Hydroxy-2-oxopropanoic acid lithium salt, 3-Hydroxy-2-oxopropionic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2COCOOLi · aq
CAS Number:
Molecular Weight:
109.99 (anhydrous basis)
Beilstein:
5774840
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (calc. based on dry substance, NT)

impurities

~1 mol water

SMILES string

[Li+].O.OCC(=O)C([O-])=O

InChI

1S/C3H4O4.Li.H2O/c4-1-2(5)3(6)7;;/h4H,1H2,(H,6,7);;1H2/q;+1;/p-1

InChI key

YHTVWBANKNBEKJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Chiral polyol synthesis catalyzed by a thermostable Transketolase immobilized on Layered Double Hydroxides in Ionic liquids: Examines the catalytic applications of lithium β-hydroxypyruvate in chiral synthesis processes (Ali et al., 2015).

Other Notes

Substrate for transketolase; Preparation of optically pure L-2-hydroxy aldehydes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O N Solov'eva et al.
Biochemistry. Biokhimiia, 66(8), 932-936 (2001-09-22)
The interaction of transketolase ketosubstrates with the holoenzyme has been studied. On addition of ketosubstrates cleaving both irreversibly (hydroxypyruvate) and reversibly (xylulose 5-phosphate), identical changes in the CD spectrum at 300-360 nm are observed. The changes in this spectral region
Shweta Joshi et al.
Molecular and biochemical parasitology, 160(1), 32-41 (2008-05-06)
The pentose phosphate pathway (PPP) is an important metabolic pathway for yielding reducing power in the form of NADPH and production of pentose sugar needed for nucleic acid synthesis. Transketolase, the key enzyme of non-oxidative arm of PPP, plays a
Yaovalak Teerajetgul et al.
Urological research, 35(4), 173-178 (2007-06-15)
We studied the effects of an intravenous hydroxypyruvate load on endogenous oxalogenesis in rats receiving a standard diet or a vitamin-B6-deficient diet. Twelve male Wistar rats were randomized to two groups and were fed either a standard diet or a
S Matosevic et al.
Journal of biotechnology, 155(3), 320-329 (2011-08-03)
Complex molecules are synthesised via a number of multi-step reactions in living cells. In this work, we describe the development of a continuous flow immobilized enzyme microreactor platform for use in evaluation of multi-step bioconversion pathways demonstrating a de novo
F. Effenberger et al.
Tetrahedron Letters, 33, 5157-5157 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service