Skip to Content
Merck
All Photos(2)

Key Documents

548644

Sigma-Aldrich

Dibenz [b,f]azepine-5-carbonyl chloride

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10ClNO
CAS Number:
Molecular Weight:
255.70
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

mp

149-153 °C (lit.)

SMILES string

ClC(=O)N1c2ccccc2C=Cc3ccccc13

InChI

1S/C15H10ClNO/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-10H

InChI key

APJYHXJGXDPGBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibenz [b,f]azepine-5-carbonyl chloride or 5H-dibenz [b,f]azepine-5-carbonyl chloride is a tricyclic heterocyclic compound that can be synthesized from 5H-dibenz[ b,f]azepine.

Application

Dibenz [b,f]azepine-5-carbonyl chloride may be used in the preparation of trans-10,11-dibromo-10,11-dihydro-5H-dibenz[b,f]azepine-5-carbonyl chloride via bromination using bromine. It may also be used to prepare urea derivatives, which are potent P2X4 receptor (purinergic receptor) antagonists.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic evidence for rate determination during the nucleophilic step of olefin bromination. The case of 5H-dibenz [b, f] azepine-5-carbonyl chloride.
Bellucci G and Chiappe C.
The Journal of Organic Chemistry, 58(25), 7120-7127 (1993)
Maoqun Tian et al.
Bioorganic & medicinal chemistry, 22(3), 1077-1088 (2014-01-15)
Antagonists for the P2 receptor subtype P2X4, an ATP-activated cation channel receptor, have potential as novel drugs for the treatment of neuropathic pain and other inflammatory diseases. In the present study, a series of 47 carbamazepine derivatives including 32 novel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service