Skip to Content
Merck
All Photos(1)

Key Documents

525138

Sigma-Aldrich

2-Fluoroethyl chloroformate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2CH2CH2F
CAS Number:
Molecular Weight:
126.51
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.83 mmHg ( 20 °C)

Assay

97%

refractive index

n20/D 1.4 (lit.)

bp

131-135 °C (lit.)

density

1.34 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FCCOC(Cl)=O

InChI

1S/C3H4ClFO2/c4-3(6)7-2-1-5/h1-2H2

InChI key

FOHSVPIFZASOED-UHFFFAOYSA-N

General description

2-Fluoroethyl chloroformate is an alkyl fluorinated building block.

Application

2-Fluoroethyl chloroformate may be used to synthesize 3-fluoroethylurethan derivatives.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.0 °F - closed cup

Flash Point(C)

58.89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Notes-Synthesis and Investigation of Organic Fluorine Compounds. XXII. The Preparation of Newer 2-Fluoroethylurethan Derivatives.
Olah G, et al.
The Journal of Organic Chemistry, 22(8), 979-980 (1957)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service