Skip to Content
Merck
All Photos(1)

Key Documents

519235

Sigma-Aldrich

4-Ethynylbenzyl alcohol

97%

Synonym(s):

4-Hydroxymethylphenylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CC6H4CH2OH
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

40-44 °C (lit.)

functional group

hydroxyl

SMILES string

OCc1ccc(cc1)C#C

InChI

1S/C9H8O/c1-2-8-3-5-9(7-10)6-4-8/h1,3-6,10H,7H2

InChI key

QCZORVSTESPHCO-UHFFFAOYSA-N

Application

  • 4-Ethynylbenzyl alcohol can be used in the synthesis of platinum-acetylide dendrimers, rotaxanes and arylacetylenes.
  • It was employed in the preparation of arylalkyne-tagged sugars for the photoinduced glycosylation of cysteine-containing peptides.
  • It can also act as a precursor to synthesize fluorescent probes via Cu(I)-catalyzed azide-alkyne cycloaddition (CuAAC) click reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A fluorogenic `click?reaction of azidoanthracene derivatives.
Xie F, et al.
Tetrahedron, 64(13), 2906-2914 (2008)
Synthesis and conformation effects of poly (phenylacetylene) s having chiral and racemic polylactide pendants.
Zhang C, et al.
Polymer International, 61(7), 1158-1162 (2012)
Susimaire P Mantoani et al.
Molecules (Basel, Switzerland), 21(2), doi:10-doi:10 (2016-02-11)
Alzheimer's disease (AD) is the most prevalent neurodegenerative disorder worldwide. Currently, the only strategy for palliative treatment of AD is to inhibit acetylcholinesterase (AChE) in order to increase the concentration of acetylcholine in the synaptic cleft. Evidence indicates that AChE
Palladium/silver-cocatalyzed tandem reactions of oxabenzonorbornadienes with substituted arylacetylenes: A simple method for the preparation of 1, 2-diarylethanones and 1, 2-diarylacetylenes.
Chen J, et al.
Organometallics, 34(17), 4318-4322 (2015)
Kideog Bae et al.
Analytical chemistry, 92(1), 740-748 (2019-11-22)
The dynamics of mitochondria in live cells play a pivotal role in biological events such as cell metabolism, early stage apoptosis, and cell differentiation. Triphenylphosphonium (TPP) is a commonly used mitochondria-targeting agent for mitochondrial studies. However, there has been a

Articles

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service