Skip to Content
Merck
All Photos(1)

Key Documents

496391

Sigma-Aldrich

1,1′-Ferrocenedicarboxaldehyde

96%

Synonym(s):

1,1′-Bisformylferrocene, 1,1′-Diformylferrocene, Cyclopentadienecarboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10FeO2
CAS Number:
Molecular Weight:
242.05
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

reaction suitability

core: iron
reagent type: catalyst

mp

151-156 °C (lit.)

SMILES string

[Fe].O=C[C]1[CH][CH][CH][CH]1.O=C[C]2[CH][CH][CH][CH]2

InChI

1S/2C6H5O.Fe/c2*7-5-6-3-1-2-4-6;/h2*1-5H;

InChI key

SRAPEJKOIJWBFN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service