All Photos(1)
About This Item
Linear Formula:
C6D5CD2CD(NH2)CO2H
CAS Number:
Molecular Weight:
173.24
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
isotopic purity
98 atom % D
Quality Level
form
solid
mp
266-267 °C (dec.) (lit.)
mass shift
M+8
SMILES string
[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])C([2H])(N)C(O)=O
InChI
1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/i1D,2D,3D,4D,5D,6D2,8D
InChI key
COLNVLDHVKWLRT-INHGFZCOSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service