Skip to Content
Merck
All Photos(1)

Key Documents

485128

Sigma-Aldrich

Benzyl alcohol, polymer-bound

100-200 mesh, extent of labeling: 1.0-2.0 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym(s):

(Hydroxymethyl)polystyrene, Benzyl alcohol polymer-bound, Hydroxymethyl polystyrene, Polystyrene cross-linked with divinylbenzene, hydroxymethylated

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.22

Quality Level

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: Boc solid-phase peptide synthesis

extent of labeling

1.0-2.0 mmol/g loading

particle size

100-200 mesh

functional group

alcohol

SMILES string

OCc1ccccc1.C=Cc2ccccc2.C=Cc3ccc(C=C)cc3

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service