Skip to Content
Merck
All Photos(1)

Documents

47633

Sigma-Aldrich

Fmoc-Leu-OH

≥97.0%

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-leucine, Fmoc-L-leucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
Beilstein:
2178254
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Assay

≥97.0%

optical activity

[α]20/D −25±2°, c = 1% in DMF

reaction suitability

reaction type: C-H Activation
reaction type: Fmoc solid-phase peptide synthesis
reagent type: ligand
reaction type: Peptide Synthesis

mp

152-156 °C (lit.)
152-156 °C

application(s)

peptide synthesis

functional group

Fmoc
amine
carboxylic acid

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C21H23NO4/c1-13(2)11-19(20(23)24)22-21(25)26-12-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h3-10,13,18-19H,11-12H2,1-2H3,(H,22,25)(H,23,24)/t19-/m0/s1

InChI key

CBPJQFCAFFNICX-IBGZPJMESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fmoc-Leu-OH can be used as a reactant to synthesize:
  • Various oligopeptides by reacting with functionalized α-amino acid hydrochloride salts.
  • A cyclic depsipeptide sansalvamide A, a natural product found in marine fungus.
  • Streptocidin A−D, decapeptide antibiotics naturally found in Streptomyces sp. Tü 6071.
  • Coumaroyl dipeptide amide that can be used for cosmetic applications.

Biochem/physiol Actions

PPARγ ligand that induces insulin sensitization, but not adipogenesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile solid-phase synthesis of cyclic decapeptide antibiotic streptocidins A-D
Qin C, et al.
Tetrahedron Letters, 45(1), 217-220 (2004)
Markus Beck Erlach et al.
Journal of biomolecular NMR, 69(2), 53-67 (2017-09-16)
For evaluating the pressure responses of folded as well as intrinsically unfolded proteins detectable by NMR spectroscopy the availability of data from well-defined model systems is indispensable. In this work we report the pressure dependence of
Synthesis and Evaluation of Coumaroyl Dipeptide Amide as Potential Whitening Agents
Lee H-S, et al.
Bull. Korean Chem. Soc., 34(10), 3017-3021 (2013)
Zhimou Yang et al.
Chemical communications (Cambridge, England), (35)(35), 4414-4416 (2005-09-02)
Two types of therapeutic agents, which have discrete yet complementary functions, self-assemble into nanofibers in water to formulate a new supramolecular hydrogel as a self-delivery biomaterial to reduce the toxicity of uranyl oxide at the wound sites.
Franziska Thomas et al.
Biochemistry, 56(50), 6544-6554 (2017-11-23)
Coiled coils (CCs) are among the best-understood protein folds. Nonetheless, there are gaps in our knowledge of CCs. Notably, CCs are likely to be structurally more dynamic than often considered. Here, we explore this in an abundant class of CCs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service