Skip to Content
Merck
All Photos(2)

Key Documents

467359

Sigma-Aldrich

(S)-(−)-4-Amino-2-hydroxybutyric acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH2CH(OH)CO2H
CAS Number:
Molecular Weight:
119.12
Beilstein:
1721686
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
EC Index Number:
670-305-7
NACRES:
NA.22

Assay

96%

optical activity

[α]23/D −30°, c = 1 in H2O

mp

200-203 °C (lit.)

SMILES string

NCC[C@H](O)C(O)=O

InChI

1S/C4H9NO3/c5-2-1-3(6)4(7)8/h3,6H,1-2,5H2,(H,7,8)/t3-/m0/s1

InChI key

IVUOMFWNDGNLBJ-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(-)-4-Amino-2-hydroxybutyric acid is an important moiety of butirosin, an aminoglycoside antibiotic.

Application

(S)-(-)-4-Amino-2-hydroxybutyric acid may be used in the preparation of 6′-amino-1-N-[(S)-4-amino-2-hydroxybutyryl]-6′-deoxylividomycin A.
Building block for enantiopure 3-hydroxypyrrolidin-2-ones.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 6'-Amino-1-N-[(S)-4-Amino-2-Hydroxybutyryl]-6'-Deoxylividomycin A.
Watanabe I, et al.
Bulletin of the Chemical Society of Japan, 48(8), 2303-2305 (1975)
Synthesis of (S)-Isoserine.
Miyazawa T, et al.
Agricultural and Biological Chemistry, 40(8), 1651-1652 (1976)
M Philippe et al.
The Journal of antibiotics, 35(11), 1507-1512 (1982-11-01)
A semisynthetic aminoglycoside antibiotic 15, containing a cyclic gamma-amino-alpha-hydroxy acid, related to the 1-N-4-amino-2-hydroxybutyric acid (AHBA) side chain of butirosins and amikacin, has been prepared. Conveniently protected 3,2',6'-tris-N-tert-butoxycarbonylgentamicin C1a (12) was condensed with the phtalimido active ester 10 to give
Bentley, J.M. et al.
Journal of the Chemical Society. Chemical Communications, 231-231 (1995)
Y Ota et al.
The Journal of antibiotics, 53(10), 1158-1167 (2001-01-02)
Butirosin is an interesting 2-deoxystreptamine (DOS)-containing aminoglycoside antibiotic produced by non-actinomycete Bacilli. Recently we were successful in purification of 2-deoxy-scyllo-inosose synthase from butirosin-producer Bacillus circulans as the key enzyme for the biosynthesis of DOS, in cloning of the responsible gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service