Skip to Content
Merck
All Photos(3)

Documents

444545

Sigma-Aldrich

Cobalt(II) 2-ethylhexanoate solution

65 wt. % in mineral spirits

Synonym(s):

2-Ethylhexanoic acid cobalt(2+) salt, Bis(2-ethylhexanoate)cobalt, Cobalt bis(2-ethylhexanoate), Cobalt octoate, Cobaltous octoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3CH(C2H5)CO2]2Co
CAS Number:
Molecular Weight:
345.34
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

composition

Co, 11.0-13.0% EDTA titration

reaction suitability

core: cobalt

concentration

65 wt. % in mineral spirits

density

1.002 g/mL at 25 °C

SMILES string

CCCCC(CC)C(=O)O[Co]OC(=O)C(CC)CCCC

InChI

1S/2C8H16O2.Co/c2*1-3-5-6-7(4-2)8(9)10;/h2*7H,3-6H2,1-2H3,(H,9,10);/q;;+2/p-2

InChI key

QAEKNCDIHIGLFI-UHFFFAOYSA-L

Application

Polymerization accelerator

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eliška Matušková et al.
Materials (Basel, Switzerland), 13(3) (2020-02-07)
This paper reports a strong drying activity of manganese(III) acetylacetonate. It is documented on several solvent-borne and high-solid alkyd binders. Solubility problems, which often appear upon application of new primary driers, were overcome by use of dimethyl sulfoxide. Interestingly, intense
Offset printer's occupational allergic contact dermatitis caused by cobalt-2-ethylhexoate.
L Kanerva et al.
Contact dermatitis, 34(1), 67-68 (1996-01-01)
Occupational cobalt-allergic contact dermatitis resulting from polyester resin.
Jennifer L Cahill et al.
Contact dermatitis, 63(5), 292-294 (2010-10-16)
Namrata S Anavekar et al.
The Australasian journal of dermatology, 47(2), 143-144 (2006-04-28)
A 46-year-old woman, who worked as a laminator of spa baths, presented with hand dermatitis, which was suspected to be related to her occupation. Patch testing revealed strong reactions to both cobalt chloride and a polyester resin that the patient
Jan Honzíček et al.
Materials (Basel, Switzerland), 14(5) (2021-04-04)
This study describes the catalytic performance of an iron(III) complex bearing a phthalocyaninato-like ligand in two solvent-borne and two high-solid alkyd binders. Standardized mechanical tests revealed strong activity, which appeared in particular cases at concentrations about one order of magnitude

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service