Skip to Content
Merck
All Photos(1)

Key Documents

440337

Sigma-Aldrich

Tung oil

Synonym(s):

China wood oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

gel time 12 min

form

liquid

composition

ester of eleostearic acid, 80%
esters of linolenic, 9,12-linoleic, oleic, stearic and palmitic acids, 20%

color

Gardner: ≤12

refractive index

n20/D 1.52 (lit.)

acid number

≤6.9 mg KOH/g

saponification value

189‑195 mg KOH/g

density

0.937 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

Tung oil is also called china wood oil that can be produced by cold pressing the seeds of tung tree. It can be used as a biodiesel and a drying oil in the production of paints.

Application

Tung oil is a plant oil that can be polymerized to produce a variety of products which include pressure sensitive adhesives, self-healed epoxy coatings, polyurethane foam, and vinyl ester resins.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

552.2 °F - closed cup

Flash Point(C)

289 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Polyurethane foams based on modified tung oil and reinforced with rice husk ash II: Mechanical characterization.
da Silva VR, et al.
Polymer Testing, 32(4), 665-672 (2013)
Tung oil: An autonomous repairing agent for self-healing epoxy coatings.
Samadzadeh M, et al.
Progress in Organic Coatings, 70(4), 383-387 (2011)
Polyurethanes from tung oil: polymer characterization and composites.
Mosiewicki MA, et al.
Polym. Eng. Sci., 49(4), 685-692 (2009)
Properties of Tung oil biodiesel and its blends with diesel.
Shang Q, et al.
Bioresource Technology, 101(2), 826-828 (2010)
Preparation of pressure-sensitive adhesives from tung oil via Diels-Alder reaction.
Gallart-Sirvent P, et al.
International Journal of Adhesion and Adhesives, 78(4), 67-73 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service