Skip to Content
Merck
All Photos(2)

Documents

427888

Sigma-Aldrich

Polypropylene

Isotactic, average Mw ~250,000, average Mn ~67,000

Synonym(s):

PP, polypropene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

beads

melt index

12 g/10 min (230°C/2.16kg)

mol wt

average Mn ~67,000
average Mw ~250,000

hardness

100 (Rockwell R, ASTM D 785-A)

transition temp

Tm 160-165 °C

density

0.9 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

CC=C

InChI

1S/C22H42O3/c1-2-3-4-5-11-14-17-20-21(25-20)18-15-12-9-7-6-8-10-13-16-19-22(23)24/h20-21H,2-19H2,1H3,(H,23,24)/t20-,21+/m1/s1

InChI key

NSYDMBURIUSUDH-RTWAWAEBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used with ram- and screw-injection machines. For automotive, housewares, general molding products and multi- and monofiliment fiber.

Features and Benefits

High stiffness and resistance to solvents and chemicals. Resistant to environmental stress cracking. More prone to oxidation than polyethylene.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Yan et al.
Food microbiology, 87, 103354-103354 (2020-01-18)
Viruses are transmissible via their interaction with contact surfaces of food containers or tools. This study evaluated the recoveries of MS2 coliphage, a virus surrogate, from polypropylene (PP), polyvinyl chloride (PVC), polyethylene (PE), and glass (borosilicate and soda lime), as
Simon J Gallagher et al.
Journal of drug targeting, 11(6), 373-379 (2003-12-12)
The aim of this study was to test the hypothesis that the most appropriate model for studying the diffusional release of an active from a topical formulation is one in which the membrane offers minimal resistance to release and involves
G Imanidis et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 47(3), 283-287 (1999-06-26)
The possibility to control the release rate of salbutamol through the hydrophobic Celgard 2500 polypropylene membrane by varying the composition and the viscosity of hydrophilic drug vehicles was investigated. The use of the polypropylene membrane as a control membrane for
F Cassiola et al.
Artificial organs, 24(3), 168-173 (2000-04-12)
The introduction of microporous polypropylene hollow fibers in recent years has brought considerable advances to blood oxygenators. However, lifetime and assembly problems are still unresolved. In this work we tried to rate the oxygen permeation velocity by turning the fibers
[Procedure for membrane oxygenation of blood over hydrophilic polymer membranes].
H Körber et al.
Biomedizinische Technik. Biomedical engineering, 43 Suppl, 338-340 (1998-12-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service