Skip to Content
Merck
All Photos(1)

Key Documents

406457

Sigma-Aldrich

Methyl perfluorooctanoate

98%

Synonym(s):

Methyl pentadecafluorooctanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)6CO2CH3
CAS Number:
Molecular Weight:
428.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.305 (lit.)

bp

159-160 °C (lit.)

density

1.786 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C9H3F15O2/c1-26-2(25)3(10,11)4(12,13)5(14,15)6(16,17)7(18,19)8(20,21)9(22,23)24/h1H3

InChI key

XOCNYZFAMHDXJK-UHFFFAOYSA-N

General description

Methyl perfluorooctanoate (MePFOA) is reported to be formed during storage of perfluorooctanoic acid (PFOA) in methanol. Its physical properties like density, boiling point and refractive index have been determined experimentally.

Application

Methyl perfluorooctanoate may be employed in the following studies:
  • Synthesis of monodisperse perfluoroalkyl N-polyethoxylated amides, potential nonionic fluorinated surfactants.
  • To investigate the mutual solubility curves of fluorocarbon-hydrocarbon systems.
  • Synthesis of fluorine-containing monomer, N-(5-hydroxypent-1-yl)perfluorooctaneamide.
  • To investigate the vinyl ether formulations for use in step and flash imprint lithography.
  • Preparation of 2-chloro-1,1-dicyano-2-(perfluoroheptyl)ethylene, a precursor for synthesizing perfluorocarbon-soluble dyes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(α-Perfluoroheptyl-β,β-dicyanovinyl) aminostyrenes: A novel class of perfluorocarbon-soluble dyes.
Freed BK and Middleton WJ.
Journal of Fluorine Chemistry, 47(2), 219-225 (1990)
Syntheses of perfluoroalkyl N-polyethoxylated amides.
Afzal J, et al.
Journal of Fluorine Chemistry, 34(3), 385-393 (1987)
Free energy relations in fluorocarbon-hydrocarbon systems.
Kyle BG and Reed III TM.
Journal of the American Chemical Society, 80(23), 6170-6177 (1958)
Variation in concentration of perfluorooctanoic acid in methanol solutions during storage.
Hanari N, et al.
Chemosphere, 94, 116-120 (2014)
Vinyl ether formulations for step and flash imprint lithography.
Kim E K, et al.
J. Vac. Sci. Technol. B, 23(6), 2967-2971 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service