Skip to Content
Merck
All Photos(1)

Key Documents

390755

Sigma-Aldrich

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate

97%

Synonym(s):

Difluoro(fluorosulfonyl)acetic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FSO2CF2CO2CH3
CAS Number:
Molecular Weight:
192.11
Beilstein:
1812896
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

reaction suitability

reaction type: click chemistry

refractive index

n20/D 1.351 (lit.)

bp

117-118 °C (lit.)

density

1.509 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(F)(F)S(F)(=O)=O

InChI

1S/C3H3F3O4S/c1-10-2(7)3(4,5)11(6,8)9/h1H3

InChI key

GQJCAQADCPTHKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Useful reagent for the trifluoromethylation of alkyl halides.
  • Broadly applicable reagent for perfluoroalkylations for aryl iodides and bromides

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Chemical Communications, 705-705 (1989)
A broadly applicable copper reagent for trifluoromethylations and perfluoroalkylations of aryl iodides and bromides.
Hiroyuki Morimoto et al.
Angewandte Chemie (International ed. in English), 50(16), 3793-3798 (2011-03-29)
David Orr et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(44), 14305-14316 (2014-09-13)
Vinyl cyclopropane rearrangement (VCPR) has been utilised to synthesise a difluorinated cyclopentene stereospecifically and under mild thermal conditions. Difluorocyclopropanation chemistry afforded ethyl 3-(1'(2'2'-difluoro-3'-phenyl)cyclopropyl) propenoate as all four stereoisomers (18a, 18b, 22a, 22b) (all racemic). The trans-E isomer (18a), prepared in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service