Skip to Content
Merck
All Photos(2)

Documents

380016

Sigma-Aldrich

Sodium thiosulfate pentahydrate

99.999% trace metals basis

Synonym(s):

Disodium thiosulfate pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O3 · 5H2O
CAS Number:
Molecular Weight:
248.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

powder

impurities

≤15.0 ppm Trace Metal Analysis

SMILES string

O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=S

InChI

1S/2Na.H2O3S2.5H2O/c;;1-5(2,3)4;;;;;/h;;(H2,1,2,3,4);5*1H2/q2*+1;;;;;;/p-2

InChI key

PODWXQQNRWNDGD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Applied in a widely useful, thiosulfate-assisted, synthesis of a variety of diamine-CuCN complexes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fred B. Stocker et al.
Inorganic chemistry, 38(5), 984-991 (2001-10-24)
A new synthetic procedure developed recently in our laboratories has made possible the synthesis of variety of new complexes of CuCN with diamines. Synthesis was effected by adding the ligand to a solution of CuCN in aqueous sodium thiosulfate. This
Melani Sooriyaarachchi et al.
Metallomics : integrated biometal science, 5(3), 197-207 (2013-02-28)
Cis-Platin (CP) is a remarkably effective Pt-based anticancer drug, but it also exhibits severe toxic side-effects, including nephrotoxicity and ototoxicity. Previous studies conducted with mammalian model organisms have clearly demonstrated that the intravenous administration of N-acetyl-L-cysteine (NAC) or sodium thiosulfate
Michael C Reade et al.
Emergency medicine Australasia : EMA, 24(3), 225-238 (2012-06-08)
Cyanide poisoning is uncommon, but generates interest because of the presumed utility of an antidote immediately available in those areas with a high risk of cyanide exposure. As part of its regular review of guidelines, the Australian Resuscitation Council conducted
Brooke Bair et al.
Journal of drugs in dermatology : JDD, 10(9), 1042-1044 (2011-11-05)
Calcinosis cutis is a term used to describe a group of disorders which result in calcium deposits in the skin. These disorders can be separated based on etiology. Sodium thiosulfate has been used to systemically treat calciphylaxis with little to
Stephanie Chiu et al.
American journal of infection control, 43(11), 1208-1212 (2015-08-10)
The efficacies of disinfection by sodium hypochlorite, accelerated hydrogen peroxide (AHP), and quaternary ammonium compound (QUAT) commonly used in health care facilities were determined using the surrogate viruses murine norovirus (MNV-1) and feline calicivirus (FCV). A virus suspension of known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service