Skip to Content
Merck
All Photos(1)

Key Documents

324647

Sigma-Aldrich

Allylboronic acid pinacol ester

97%

Synonym(s):

2-Allyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-(2-propen-1-yl)-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-(2-propenyl)-1,3,2-dioxaborolane, Allyl pinacol boronate, Pinacol allylboronate, Pinacolyl 2-propenylboronate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)4C2O2)BCH2CH=CH2
CAS Number:
Molecular Weight:
168.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.4268 (lit.)

bp

50-53 °C/5 mmHg (lit.)

density

0.896 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)OB(CC=C)OC1(C)C

InChI

1S/C9H17BO2/c1-6-7-10-11-8(2,3)9(4,5)12-10/h6H,1,7H2,2-5H3

InChI key

YMHIEPNFCBNQQU-UHFFFAOYSA-N

General description

Allylboronic acid pinacol ester is used as a nucleophile in the catalytic allylation of simple ketoimines.

Application

Reagent used for
  • Palladium-catalyzed Suzuki-Miyaura cross-coupling reactions and olefin metathesis
  • Intermolecular radical additions
  • Allylboration of aldehydes catalyzed by chiral spirobiindane diol (SPINOL) based phosphoric acids
  • Cobalt-catalyzed regioselective hydrovinylation of dienes with alkenes
  • Nucleic acid-templated energy transfer leading to a photorelease reaction
  • Stereoselective indium-catalyzed Hosomi-Sakurai reactions

Reagent used in Preparation of
  • Cyclic sulfone hydroxyethylamines as BACE1 inhibitors for reduction of Amyloid β-Peptides
  • Transmetalation of carbene Ru iodide with Ag carboxylates to give C-H-activated Ru carbene complexes as catalysts for Z-selective olefin metathesis
  • Allylboronation reagent for the preparation of allylic alcohols, and homoallylic amines.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marion Arndt et al.
Organic letters, 13(23), 6236-6239 (2011-11-02)
The absolute control of the regiochemistry of a cobalt-catalyzed 1,4-hydrovinylation reaction is achieved by alternation of the ligands applied. While the dppe/dppp ligands led to the formation of the branched product, the herein described application of the SchmalzPhos ligand generates
Organometallics, 2, 230-230 (1983)
Asymmetric Allylboration of Aldehydes with Pinacol Allylboronates Catalyzed by 1,1'-Spirobiindane-7,7'-diol (SPINOL) Based Phosphoric Acids
Xing, C-H.; et al.
European Journal of Organic Chemistry, 6, 1115-1118 (2012)
A catalytic asymmetric borono variant of Hosomi-Sakurai reactions with N,O-aminals.
Yi-Yong Huang et al.
Angewandte Chemie (International ed. in English), 50(47), 11121-11124 (2011-10-07)
Sambasivarao Kotha et al.
The Journal of organic chemistry, 77(1), 482-489 (2011-12-02)
Palladium-catalyzed Suzuki-Miyaura (SM) cross-coupling reaction with allylboronic acid pinacol ester and titanium assisted cross-metathesis (CM)/ring-closing metathesis (RCM) cascade has been used to synthesize macrocyclic cyclophane derivatives.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service