291145
N-Ethylmethylamine
97%
Synonym(s):
N-Methylethylamine, NMEA
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
C2H5NHCH3
CAS Number:
Molecular Weight:
59.11
Beilstein:
1730786
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
8.53 psi ( 20 °C)
Quality Level
Assay
97%
refractive index
n20/D 1.374 (lit.)
bp
36-37 °C (lit.)
density
0.688 g/mL at 25 °C (lit.)
functional group
amine
storage temp.
2-8°C
SMILES string
CCNC
InChI
1S/C3H9N/c1-3-4-2/h4H,3H2,1-2H3
InChI key
LIWAQLJGPBVORC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
The kinetics of reaction of guanosine 5′-phospho-2-methylimidazolide with N-ethylmethylamine has been studied in water at 37°C.
Application
N-Ethylmethylamine has been used in the synthesis of N-ethylmethylacrylamide via reaction with acryloyl chloride.
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1A
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
<-29.2 °F - closed cup
Flash Point(C)
< -34 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis of low-polydispersity poly (N-ethylmethylacrylamide) by controlled radical polymerizations and their thermal phase transition behavior.
Xu J, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 46(1), 60-69 (2008)
Zhigang Li et al.
Chemosphere, 212, 872-880 (2018-09-09)
In this study, the molecularly imprinted polymers (MIPs) with high specific surface area and extraction efficiency of N-Nitrosodiphenylamine (NDPhA) were successfully prepared and a highly sensitive and selective method was developed for determination of NDPhA in water samples using MIPs
Yuesheng Qiu et al.
Food chemistry, 232, 763-769 (2017-05-12)
The QuEChERS sample preparation method and gas chromatography-tandem mass spectrometry were employed to determine nine volatile N-nitrosamines (VNAs) in Chinese salted fish. This method was validated by considering calibration plot linearity, selectivity, matrix effects, trueness, precision, limits of quantification and
Xianze Wang et al.
Journal of environmental sciences (China), 50, 65-71 (2016-12-31)
The presence of mutagenic and carcinogenic nitrosamines in water is of great concern. In this study, seven nitrosamines including N-nitrosodimethylamine (NDMA), N-nitrosodiethylamine (NDEA), N-nitrosomethylethylamine (NMEA), N-nitrosopyrrolidine (NPyr), N-nitrosopiperidine (NPip), N-nitrosodi-n-propylamine (NDPA), and N-nitrosodi-n-butyl-amine (NDBA) were investigated in river water and
Sidsel D Schrøder et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 173, 201-206 (2016-11-05)
Absolute intensities of NH-stretching fundamental and overtone transitions of gas phase aniline, methylamine, ethylamine, cyclopropylamine, methylethylamine, diethylamine and pyrrolidine have been measured with long path length conventional absorption spectroscopy. To support the assignments of NH-stretching transitions, transition frequencies and intensities
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service