Skip to Content
Merck
All Photos(2)

Key Documents

261394

Sigma-Aldrich

1,1-Dimethylurea

99%

Synonym(s):

N,N-Dimethylurea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCONH2
CAS Number:
Molecular Weight:
88.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

178-183 °C (lit.)

solubility

water: soluble 5%, clear, colorless

SMILES string

CN(C)C(N)=O

InChI

1S/C3H8N2O/c1-5(2)3(4)6/h1-2H3,(H2,4,6)

InChI key

YBBLOADPFWKNGS-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

General description

Nonlinear optical properties of 1,1-dimethylurea (N,N′ dimethylurea), have been evaluated through second-harmonic generation.

Application

1,1-Dimethylurea (N,N-dimethylurea) has been used in the Dowex-50W ion exchange resin-promoted synthesis of N,N′-disubstituted-4-aryl-3,4-dihydropyrimidinones.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanli Lu et al.
Chemistry & biology, 18(4), 425-431 (2011-04-26)
Pactamycin, one of the most densely functionalized aminocyclitol antibiotics, has pronounced antibacterial, antitumor, antiviral, and antiplasmodial activities, but its development as a clinical drug was hampered by its broad cytotoxicity. Efforts to modulate the biological activity by structural modifications using
K Yamada et al.
Acta endocrinologica, 128(4), 379-384 (1993-04-01)
We investigated the effect of free radical scavengers on the actions of cytokines on islet cells. Interferon-gamma and tumor necrosis factor-alpha reduced the nicotinamide adenine dinucleotide content of mouse islet cells; the combination of interferon-gamma (4 x 10(5) U/l) and
S Sandler et al.
Endocrinology, 124(3), 1492-1501 (1989-03-01)
This study aimed at a more detailed characterization of the mechanisms by which interleukin 1 (IL-1) inhibits insulin secretion. For this purpose, isolated rat pancreatic islets were kept in tissue culture for 5 days in medium RPMI 1640 plus 10%
G P Meshram et al.
Mutation research, 279(4), 275-280 (1992-06-16)
Methyl isocyanate (MIC) in aqueous solution forms methylamine (MA) and N,N'-dimethylurea (DMU). MA in buffered system further converts into its salt form, methylamine hydrochloride (MAH). Therefore, MAH and DMU were evaluated for their mutagenic activity in the in vitro Ames
J W McDonald et al.
The American review of respiratory disease, 143(4 Pt 1), 806-809 (1991-04-01)
Eugenol, an extract of cloves, has been associated with pulmonary edema when inhaled from commercially available clove cigarettes. We tested the hypothesis that eugenol directly causes lung edema through oxidant-mediated mechanisms by infusing eugenol (0.1 and 1.0 mM) into isolated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service