Skip to Content
Merck
All Photos(2)

Key Documents

260916

Sigma-Aldrich

Ethylenebis(triphenylphosphine)platinum(0)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]2Pt(H2C=CH2)
CAS Number:
Molecular Weight:
747.70
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: platinum
reagent type: catalyst

mp

129-132 °C (dec.) (lit.)

SMILES string

[Pt].C=C.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6

InChI

1S/2C18H15P.C2H4.Pt/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2;/h2*1-15H;1-2H2;

InChI key

RZUASTIKPBCXPU-UHFFFAOYSA-N

Application

Nucleophilic metal species for ligand exchangewith alkenes and alkynes; catalysts for hydrosilation; reagent for oxidative addition to activated C-X bonds

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kurosawa, H.; Ogoshi, S. et al.
Journal of the American Chemical Society, 112, 2813-2813 (1990)
Gevorgyan, V.; Borisova, L. et al.
Journal of Organometallic Chemistry, 424, 15-15 (1992)
Uchimaru,Y.; Brandl, P.; Tanaka, M.; Goto, M.
Journal of the Chemical Society. Chemical Communications, 744-744 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service