Skip to Content
Merck
All Photos(1)

Key Documents

244929

Sigma-Aldrich

Dichloro(ethylenediamine)platinum(II)

99%

Synonym(s):

Dichloro(1,2-diaminoethane)platinum, Dichloro(1,2-ethanediamine)platinum, Platinum ethylenediamine dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2NCH2CH2NH2)PtCl2
CAS Number:
Molecular Weight:
326.08
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

reaction suitability

core: platinum
reagent type: catalyst

SMILES string

Cl[Pt]Cl.NCCN

InChI

1S/C2H8N2.2ClH.Pt/c3-1-2-4;;;/h1-4H2;2*1H;/q;;;+2/p-2

InChI key

LMABILRJNNFCPG-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Kimura et al.
Cancer chemotherapy and pharmacology, 32(6), 419-424 (1993-01-01)
Expression of the heat-shock protein HSP-60 is associated with poor survival in patients with ovarian carcinoma. We examined both the nature of the interaction between hyperthermia and cisplatin (DDP) using the human ovarian carcinoma cell line 2008 and the effect
T J Kelley et al.
Cancer biochemistry biophysics, 13(3), 135-146 (1993-06-01)
Cisplatin (cis-diamminedichloroplatinum(II); cis-DDP) is used as an effective drug for treatment of a variety of cancers, such as carcinomas of bladder, ovarian, and testicular origin. Immunopurified DNA polymerase-alpha from rat prostate tumor PA-3 cells was inhibited (50%) in the presence
B E Bowler et al.
Biochemistry, 25(10), 3031-3038 (1986-05-20)
We report the DNA binding site preferences of the novel molecule AO-Pt, in which the anticancer drug dichloro(ethylenediamine)platinum(II) is linked by a hexamethylene chain to acridine orange. The sequence specificity of platinum binding was mapped by exonuclease III digestion of
W L Armarego et al.
European journal of biochemistry, 164(2), 403-409 (1987-04-15)
Potassium tetrachloroplatinate (K2PtCl4) inactivates dihydropteridine reductase from human brain in a time-dependent and irreversible manner. The inactivation has been followed by measuring enzyme activity and fluorescence changes. The enzyme is completely protected from inactivation by NADH, the pterin cofactor [quinonoid
R J Holmes et al.
Journal of inorganic biochemistry, 85(2-3), 209-217 (2001-06-19)
A series of intercalator-tethered platinum(II) complexes PtLCl(2) have been prepared where L are the diamine ligands N-[2-[(aminoethyl)amino]ethyl]-9-aminoacridine-4-carboxamide, N-[3-[(2-aminoethyl)amino]propyl]-9-aminoacridine-4-carboxamide, N-[4-[(2-aminoethyl)amino]butyl]-9-aminoacridine-4-carboxamide and N-[5-[(aminoethyl)amino]pentyl]-9-aminoacridine-4-carboxamide and N-[6-[(aminoethyl)amino]hexyl]-9-aminoacridine-4-carboxamide. The activity of the complexes was assessed in the CH-1, CH-1cisR, 41M, 41McisR and SKOV-3 cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service