Skip to Content
Merck
All Photos(1)

Documents

235512

Sigma-Aldrich

Diethylmalonyl dichloride

98%

Synonym(s):

Diethylmalonic acid dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOC(C2H5)2COCl
CAS Number:
Molecular Weight:
197.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.459 (lit.)

bp

197-199 °C (lit.)

density

1.145 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCC(CC)(C(Cl)=O)C(Cl)=O

InChI

1S/C7H10Cl2O2/c1-3-7(4-2,5(8)10)6(9)11/h3-4H2,1-2H3

InChI key

FWKCXFPQSXNCBW-UHFFFAOYSA-N

Application

Diethylmalonyl dichloride has been used in the preparation of:
  • 5,5-diethyl-1,3-dihydroxybarbituric acid (N,N′-dihydroxyveronal) via condensation with 1,3-dibenzyloxyurea
  • 1,11-dibronzo-4,8-diaza-6,6-diethyl-5,7-dioxoundecane
  • amide containing crowned derivatives of dithiomaleonitrile

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Macrocyclic dithiomaleonitrile derivatives containing sulfur and nitrogen heteroatoms.
Lange SJ, et al.
Tetrahedron, 51(30), 8175-8188 (1995)
Pyrimidine N-oxides. IV. The N, N'-dihydroxy derivatives of phenobarbital and veronal.
Cowden W and Jacobsen NW.
Australian Journal of Chemistry, 35(4), 795-797 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service