Skip to Content
Merck
All Photos(1)

Documents

235377

Sigma-Aldrich

Butyl methyl ether

99%

Synonym(s):

α-Methoxybutane, 1-Methoxybutane, Methyl butyl ether, Methyl n-butyl ether, n-Butyl methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
1696931
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

139 mmHg ( 25 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.374 (lit.)

bp

70-71 °C (lit.)

mp

−115 °C (lit.)

density

0.744 g/mL at 25 °C (lit.)

SMILES string

CCCCOC

InChI

1S/C5H12O/c1-3-4-5-6-2/h3-5H2,1-2H3

InChI key

CXBDYQVECUFKRK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gas-phase reaction of the OH radical with butyl methyl ether in the presence of NO has been investigated by GC and in situ atmospheric pressure ionization tandem mass spectrometry.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Products of the gas-phase reactions of the OH radical with n-butyl methyl ether and 2-isopropoxyethanol: Reactions of ROC (?)< radicals.
Aschmann SM and Atkinson R.
International Journal of Chemical Kinetics, 31(7), 501-513 (1999)
Thu Minh Thi Tran et al.
Journal of chromatography. A, 1601, 274-287 (2019-06-20)
Electrospray mass spectrometry-profiling guided the metabolome investigation of a C18 reversed phase adsorbate of Opuntia stricta var. dillenii fruits following analytical, and semi-preparative high-performance countercurrent chromatography (HPCCC) fractionation, and visualization of molecular weight elution profiles based on selected single ion-traces.
Muhamad Roddy Ramli et al.
Journal of oleo science, 69(8), 815-824 (2020-07-10)
Approximately 900 tonne of crude palm oil (CPO) underwent washing using 5 to 10% hot water (90 to 95°C) at a palm oil mill. The aim of the CPO washing was to eliminate and/or reduce total chlorine content present in
Hans-Georg Schmarr et al.
Journal of chromatography. A, 1516, 135-141 (2017-08-22)
The chiral lactone 5-butyl-4-methyloxolan-2-one or 5-butyl-4-methyldihydro-2(3H)-furanone, often named whisky lactone, is found in oak wood, then contributing to the appreciated flavor of beverages stored in such wooden barrels. Its next higher homologue is named cognac lactone (5-pentyl-4-methyloxolan-2-one or 5-pentyl-4-methyldihydro-2(3H)-furanone), however
Tomoko Asai et al.
Journal of agricultural and food chemistry, 67(10), 2831-2838 (2019-02-21)
Objectives of the present study were to evaluate amounts of collagen in Japanese daily dishes and contents of food-derived collagen peptides in human blood. The meat in one serving of most Japanese daily dishes contains 0.2-2.5 g of collagen, except

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service