Skip to Content
Merck
All Photos(2)

Key Documents

219126

Sigma-Aldrich

β-Butyrolactone

98%

Synonym(s):

beta-Butyrolactone, β-Methyl-β-propiolactone, 3-Hydroxybutyric acid β-lactone, 4-Methyl-2-oxetanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O2
CAS Number:
Molecular Weight:
86.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.411 (lit.)

bp

71-73 °C/29 mmHg (lit.)

mp

−43.5 °C (lit.)

density

1.056 g/mL at 25 °C (lit.)

SMILES string

CC1CC(=O)O1

InChI

1S/C4H6O2/c1-3-2-4(5)6-3/h3H,2H2,1H3

InChI key

GSCLMSFRWBPUSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Butyrolactone undergoes ring opening polymerization in the presence of ethoxy-bridged dinuclear indium catalyst to yield biodegradable polyester poly(hydroxybutyrate). Polymerization of racemic β-butyrolactone in the presence of chiral initiators has been reported. It is versatile building block for organic synthesis.

Application

β-Butyrolactone was used in the preparation of (3-O-[oligo-(3-hydroxybutyrate ester)] fluorescein, fluorescein derivative of poly(3-hydroxybutyrate) via anionic polymerization.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly active, productive, and syndiospecific yttrium initiators for the polymerization of racemic beta-butyrolactone.
Abderramane Amgoune et al.
Angewandte Chemie (International ed. in English), 45(17), 2782-2784 (2006-03-21)
K Hemminki
Chemico-biological interactions, 34(3), 323-331 (1981-03-15)
Reactivity of beta-propiolactone, beta-butyrolactone and gamma-butyrolactone with guanosine, RNA, DNA and 4-(p-nitrobenzyl)pyridine was studied. beta-Propiolactone was 50--100 times more reactive with all the nucleophiles than beta-butyrolactone whereas gamma-butyrolactone was completely inactive. The rate of alkylation by the lactones was guanosine
J T Lee et al.
The Journal of organic chemistry, 66(16), 5424-5426 (2001-08-04)
The PPNCo(CO)(4) and BF(3) x Et(2)O catalyzed carbonylation of simple and functionalized epoxides in DME gives the corresponding beta-lactones regioselectively in good to high yields. The carbonylation occurred selectively at the unsubstituted C-O bond of the epoxide ring, and this
Lee R Rieth et al.
Journal of the American Chemical Society, 124(51), 15239-15248 (2002-12-19)
Polymerization of beta-butyrolactone (BBL) and beta-valerolactone (BVL) using the zinc alkoxide initiator (BDI-1)ZnO(i)()Pr [(BDI-1) = 2-((2,6-diisopropylphenyl)amido)-4-((2,6-diisopropylphenyl)imino)-2-pentene] proceeds very rapidly under mild conditions to produce poly(3-hydroxybutyrate) (PHB) and poly(3-hydroxyvalerate) (PHV), respectively. For the monomer-to-initiator ratio 200:1, PHB number-average molecular weights (M(n))
Stereoelective Polymerization of β-butyrolactone.
Le Borgne A and Spassky A.
Polymer, 30(12), 2312-2319 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service