Skip to Content
Merck
All Photos(3)

Key Documents

216194

Sigma-Aldrich

Hydrazine monohydrochloride

97%

Synonym(s):

Hydrazinium chloride, Hydrazinium monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · HCl
CAS Number:
Molecular Weight:
68.51
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

89 °C (lit.)

functional group

hydrazine

SMILES string

Cl[H].NN

InChI

1S/ClH.H4N2/c;1-2/h1H;1-2H2

InChI key

BIVUUOPIAYRCAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrazine monohydrochloride is commonly used as a reagent in the synthesis of pyrazoles.

Application

Hydrazine monohydrochloride has been used:
  • as catalyst in the synthesis of titanium oxide polymers via catalytic sol-gel process
  • in fabrication of amorphous titanium dioxide (TiO2) thin films exhibiting high refractive indices and high transparency
  • in preparation of ethyl 3-ethoxy-1H-pyrazole-4-carboxylate

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomáš Taubner et al.
International journal of biological macromolecules, 103, 202-207 (2017-05-21)
Alginic acid is a suitable material for modification to prepare new derivatives because of presence of its carboxyl groups. The high content of carboxyl groups over the entire length of its chain renders it an easily modifiable material with a
Sandrine Guillou et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(15), 4669-4677 (2010-03-25)
Our program, which has focused on the preparation of new pyrazole derivatives, has led us to report here an original and simplified preparation of ethyl 3-ethoxy-1H-pyrazole-4-carboxylate. This is based on the reaction of hydrazine monohydrochloride and diethyl 2-(ethoxymethylene)malonate. Further transformations
An efficient synthesis of substituted pyrazoles from one-pot reaction of ketones, aldehydes, and hydrazine monohydrochloride
V Lellek, et al.
Synlett, 29, 1071-1075 (2018)
Wataru Shimizu et al.
Journal of nanoscience and nanotechnology, 12(5), 3732-3738 (2012-08-03)
Titanium oxide polymers having a low-fractal dimension (d(f) < 2) were rapidly synthesized from titanium tetra-n-butoxide via a catalytic sol-gel process with a hydrazine monohydrochloride catalyst. Different from conventional sol-gel processes aimed at producing low-fractal dimension titanium oxide polymers, the
Ahmed M Elbedwehy et al.
Polymers, 11(12) (2019-11-30)
Super adsorbent polymers can be considered to be a very efficient solution for wastewater treatment. In general, their adsorption capacities depend on the type and amount of the functional groups present on the surface of the polymers, while their economic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service