Skip to Content
Merck
All Photos(2)

Key Documents

196487

Sigma-Aldrich

4-Biphenylacetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CH2CO2H
CAS Number:
Molecular Weight:
212.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

159-160 °C (lit.)

solubility

DMSO: soluble 50 mg/mL, clear, colorless to yellow

SMILES string

OC(=O)Cc1ccc(cc1)-c2ccccc2

InChI

1S/C14H12O2/c15-14(16)10-11-6-8-13(9-7-11)12-4-2-1-3-5-12/h1-9H,10H2,(H,15,16)

InChI key

QRZAKQDHEVVFRX-UHFFFAOYSA-N

Gene Information

human ... BAD(572)

Looking for similar products? Visit Product Comparison Guide

General description

4-Biphenylacetic acid is a potential non-steroidal antiinflammatory agent and forms solid inclusion complex with β-cyclodextrin. 4-Biphenylacetic acid on interaction with quinolone antibacterial agents induces functional blockade of the γ-aminobutyric acid receptors.

Application

4-Biphenylacetic acid was used in the synthesis of gastrosparing non-steroidal antiinflammatory drug.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Yi Mok et al.
Journal of medicinal chemistry, 56(5), 1843-1852 (2013-02-05)
β-Secretase (BACE1), the enzyme responsible for the first and rate-limiting step in the production of amyloid-β peptides, is an attractive target for the treatment of Alzheimer's disease. In this study, we report the application of the de novo fragment-based molecular
Ananda Sarkar et al.
Journal of molecular modeling, 15(10), 1221-1228 (2009-03-07)
Cyclooxygenase (COX) is a well-known enzyme, which converts arachidonic acid to prostaglandins H(2) (PGH(2)), which are the effective mediators of inflammation. 4', 5 di-substituted 3-biphenyl acetic acids (BPA) and several alpha-methyl derivatives (MBPA) of it are widely used as powerful
R F Halliwell et al.
Neuropharmacology, 43(4), 778-787 (2002-10-09)
A hybrid molecule composed of the antimicrobial, norfloxacin, linked to the non-steroidal anti-inflammatory drug (NSAID), biphenylacetic acid, which we have termed norbiphen, is a lethal convulsant in vivo and an antagonist of rodent GABA(A) receptors in vitro. In the present
Liqing Zang et al.
Zebrafish, 8(4), 203-210 (2011-12-21)
A novel protocol using gluten as a carrier material was developed to administer chemicals to adult zebrafish, per os (p.o.). To evaluate the capacity of gluten to retain chemicals, we prepared gluten granules containing eight types of chemicals with different
Samuel R Pygall et al.
International journal of pharmaceutics, 370(1-2), 110-120 (2008-12-23)
Few studies report the effects of alkalizing buffers in HPMC matrices. These agents are incorporated to provide micro-environmental buffering, protection of acid-labile ingredients, or pH-independent release of weak acid drugs. In this study, the influence of sodium citrate on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service