Skip to Content
Merck
All Photos(3)

Key Documents

186155

Sigma-Aldrich

Lithium acetylide, ethylenediamine complex

90%

Synonym(s):

Lithium acetylide ethylenediamine complex, Lithium acetylide, compd. with 1,2-ethanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiC≡CH · NH2CH2CH2NH2
CAS Number:
Molecular Weight:
92.07
Beilstein:
3593824
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90%

bp

110.6 °C (lit.)

mp

76 °C (dec.)

functional group

amine

storage temp.

2-8°C

SMILES string

[Li]C#C.NCCN

InChI

1S/C2H8N2.C2H.Li/c3-1-2-4;1-2;/h1-4H2;1H;

InChI key

QJQWXTYPTBEPGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Lithium acetylide, ethylenediamine complex is a general reagent to synthesize ethynylated ketones.
  • It is used as a ligand in the coordination chemistry to synthesize transition metal complexes such as 1-alkynyl-dimethyl(triorganophosphine)gold(III) complex.
  • It is employed in the ring opening reaction of epoxides, as in the total synthesis of (−)-goniotrionin and englerin A.
  • It can also be used in the ethynylation of alkyl halides to prepare terminal alkynes.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium Acetylide
Midland M M.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2001)
Lithium Acetylide
Midland M M.
e-EROS Encyclopedia of Reagents for Organic Synthesis (2001)
Total Synthesis of (?)-Goniotrionin.
Dias L C and Ferreira M A
The Journal of Organic Chemistry, 77(8), 4046-4062 (2012)
Preparation, structure and decomposition of gold (I) and gold (III) acetylide complexes.
Schuster O, et al.
Inorgorganica Chimica Acta, 358(5), 1429-1441 (2005)
A formal synthesis of (?)-englerin A by relay ring closing metathesis and transannular etherification.
Lee J and Parker K A
Organic Letters, 14(11), 2682-2685 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service