Skip to Content
Merck
All Photos(1)

Key Documents

184799

Sigma-Aldrich

5-Nitro-m-xylene-α,α′-diol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(CH2OH)2
CAS Number:
Molecular Weight:
183.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

94-96 °C (lit.)

SMILES string

OCc1cc(CO)cc(c1)[N+]([O-])=O

InChI

1S/C8H9NO4/c10-4-6-1-7(5-11)3-8(2-6)9(12)13/h1-3,10-11H,4-5H2

InChI key

FTLFITNFXXERLN-UHFFFAOYSA-N

Related Categories

Application

5-Nitro-m-xylene-α,α′-diol was used to fabricate chemical sensor for detection of explosive 2,4,6-trinitrotoluene by planar integrated optical waveguide attenuated total reflection spectrometry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalie R Walker et al.
Analytica chimica acta, 593(1), 82-91 (2007-05-29)
A chemical sensor was developed to detect the explosive 2,4,6-trinitrotoluene (TNT) utilizing planar integrated optical waveguide (IOW) attenuated total reflection spectrometry. Submicron thick films of organically modified sol-gel polymers were deposited on the waveguide surface as the sensing layer. Sol-gels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service