Skip to Content
Merck
All Photos(1)

Key Documents

159646

Sigma-Aldrich

N,N′-Diphenylformamidine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCH=NC6H5
CAS Number:
Molecular Weight:
196.25
Beilstein:
511953
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

136-139 °C (lit.)

storage temp.

2-8°C

SMILES string

N(\C=N\c1ccccc1)c2ccccc2

InChI

1S/C13H12N2/c1-3-7-12(8-4-1)14-11-15-13-9-5-2-6-10-13/h1-11H,(H,14,15)

InChI key

ZQUVDXMUKIVNOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N′-Diphenylformamidine is an N,N-disubstituted formamide and it forms adducts with phosgeneand oxalyl chloride.

Application

N,N′-Diphenylformamidine was used in the microwave-assisted synthesis of 5-aminopyrazol-4-yl ketones. It was also used in the preparation of monomethine dyes containing imide functional groups.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frank Würthner et al.
The Journal of organic chemistry, 68(23), 8943-8949 (2003-11-08)
The condensation of the CH acidic heterocycles 4-alkyl-2,6-dioxo-1,2,5,6-tetrahydropyridine-3-carbonitrile (5a and b) and barbituric acid (15) with electron-rich thiophene aldehydes and benzaldehyde derivatives affords the respective monomethine dyes 10-13 and 17-19. The formylation of 5a,b and 15 with N,N'-diphenylformamidine or dibutylformamide
Chemistry of the adducts of N,N'-diphenylformamidine with oxalyl chloride and phosgene.
Barsa EA and Richter R.
The Journal of Organic Chemistry, 51(23), 4483-4485 (1986)
Mark C Bagley et al.
Bioorganic & medicinal chemistry letters, 18(13), 3745-3748 (2008-06-10)
5-Aminopyrazol-4-yl ketones are prepared rapidly and efficiently using microwave dielectric heating from beta-ketonitriles by treatment with N,N'-diphenylformamidine followed by heterocyclocondensation by irradiation with a hydrazine. The inhibitory activity of RO3201195 prepared by this methodology was confirmed in hTERT-immortalized HCA2 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service