Skip to Content
Merck
All Photos(3)

Key Documents

133027

Sigma-Aldrich

Bisphenol A

97%

Synonym(s):

2,2-Bis(4-hydroxyphenyl)propane, 4,4′-Isopropylidenediphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(C6H4OH)2
CAS Number:
Molecular Weight:
228.29
Beilstein:
1107700
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

solid

bp

220 °C/4 mmHg (lit.)

mp

158-159 °C (lit.)

SMILES string

CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

InChI

1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3

InChI key

IISBACLAFKSPIT-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

440.6 °F - closed cup

Flash Point(C)

227 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Damaskini Valvi et al.
Epidemiology (Cambridge, Mass.), 24(6), 791-799 (2013-09-17)
Increasing experimental evidence suggests that prenatal bisphenol A (BPA) exposure induces offspring weight gain, but these effects remain largely unexplored in humans. We examined the effects of prenatal BPA exposure on postnatal growth and obesity. BPA concentrations were measured in
Tinne Geens et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(10), 3725-3740 (2012-08-15)
Due to the large number of applications of bisphenol-A (BPA), the human exposure routes are multiple. We aimed to review shortly the food and non-food sources of BPA, and to evaluate their contribution to the human exposure. Food sources discussed
Marie Tohmé et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(7), 3124-3133 (2014-04-20)
Bisphenol A (BPA) is an endocrine disruptor that displays estrogenic activity. Several reports suggest that BPA may have estrogen receptor-independent effects. In zebrafish, 50 μM BPA exposure induces otic vesicle abnormalities, including otolith aggregation. The purpose of this study was
R E FitzGerald et al.
Toxicology letters, 230(2), 368-374 (2014-05-17)
Bisphenol A (BPA) is one of the most widely used and extensively studied chemicals. Numerous studies have reported in vitro effects or animal adverse findings at BPA doses lower than the no observed adverse effect levels (NOAELs) established in regulatory
Albert A Koelmans et al.
Environmental pollution (Barking, Essex : 1987), 187, 49-54 (2014-01-21)
It is often assumed that ingestion of microplastics by aquatic species leads to increased exposure to plastic additives. However, experimental data or model based evidence is lacking. Here we assess the potential of leaching of nonylphenol (NP) and bisphenol A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service