Skip to Content
Merck
All Photos(2)

Documents

132195

Sigma-Aldrich

Trimethyl phosphate

97%

Synonym(s):

TMP, TMPA, TMPO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3PO
CAS Number:
Molecular Weight:
140.07
Beilstein:
1071731
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.395 (lit.)

bp

197 °C (lit.)

mp

−46 °C (lit.)

density

1.197 g/mL at 25 °C (lit.)

SMILES string

COP(=O)(OC)OC

InChI

1S/C3H9O4P/c1-5-8(4,6-2)7-3/h1-3H3

InChI key

WVLBCYQITXONBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethyl phosphate is an excellent dipolar aprotic solvent that is used as a methylating agent and acid scavenger in many organic transformations.

Application

Trimethyl phosphate can be used as a methylating agent for the:
  • Solvent-free o-methylation of phenolic compounds to synthesize methyl aryl ethers.
  • S-methylation of aryl/alkyl thiols to synthesize S-methylated product in the presence of Ca(OH)2 as a base.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongqing Ma et al.
Analytica chimica acta, 786, 47-53 (2013-06-26)
An analytical method for the determination of 14 organophosphorus flame retardants (OPFRs), including halogenated OPFRs, non-halogenated OPFRs and triphenyl phosphine oxide (TPPO) in biological samples was developed using gas chromatography-mass spectrometry (GC/MS). Biological samples were extracted using microwave-assisted extraction (MAE)
Hydrolysis of acetyl dimethyl phosphate, a reactive acyl phosphate.
R Kluger et al.
Biochemistry, 11(8), 1544-1546 (1972-04-11)
R V Banerjee et al.
Biochemistry, 27(25), 9062-9070 (1988-12-13)
The transfer of 17O and/or 18O from (COOH-17O or -18O) enriched substrates to inorganic phosphate (Pi) has been demonstrated for two enzyme-catalyzed reactions involved in folate biosynthesis and glutamylation. COOH-18O-labeled folate, methotrexate, and dihydropteroate, in addition to [17O]-glutamate, were synthesized
K Sundararajan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56A(10), 1855-1867 (2000-09-16)
Trimethyl phosphate (TMP) and acetylene were codeposited in nitrogen and argon matrices and adducts of these species were identified using infrared spectroscopy. Formation of the adducts was evidenced by shifts in the vibrational frequencies of the modes involving the TMP
Johannes Bernarding et al.
Journal of the American Chemical Society, 128(3), 714-715 (2006-01-19)
In ultralow magnetic fields, liquid state nuclear magnetic resonance (NMR) spectra of homonuclear spin systems exhibit line widths dominated by their natural lifetime. Chemical shifts become negligible, and heteronuclear NMR spectra show predominantly the electron-mediated field-independent J-coupling. However, weak polarization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service