Skip to Content
Merck
All Photos(2)

Documents

129887

Sigma-Aldrich

N-Methylisatoic anhydride

technical, 90%

Synonym(s):

1-Methyl-1H -benzo[d ][1,3]oxazine-2,4-dione, 1-Methyl-4H -3,1-benzoxazine-2,4(1H )dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO3
CAS Number:
Molecular Weight:
177.16
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

6.1 (vs air)

grade

technical

Assay

90%

mp

165 °C (dec.) (lit.)

SMILES string

CN1C(=O)OC(=O)c2ccccc12

InChI

1S/C9H7NO3/c1-10-7-5-3-2-4-6(7)8(11)13-9(10)12/h2-5H,1H3

InChI key

KJMRWDHBVCNLTQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erich Smith-Peter et al.
RNA biology, 12(12), 1372-1382 (2015-09-26)
Riboswitches regulate gene expression by rearranging their structure upon metabolite binding. The lysine-sensing lysC riboswitch is a rare example of an RNA aptamer organized around a 5-way helical junction in which ligand binding is performed exclusively through nucleotides located at
Joshua D Podlevsky et al.
RNA (New York, N.Y.), 22(2), 204-215 (2015-11-26)
Telomerase is a ribonucleoprotein (RNP) enzyme that requires an integral telomerase RNA (TR) subunit, in addition to the catalytic telomerase reverse transcriptase (TERT), for enzymatic function. The secondary structures of TRs from the three major groups of species, ciliates, fungi
G Knipping et al.
The International journal of biochemistry, 25(4), 505-511 (1993-04-01)
1. To investigate whether a direct protein-protein interaction between apoA-I and lecithin:cholesterol acyltransferase (LCAT) is necessary for the activation of the enzyme, apoA-I was labelled with N-methylisatoic anhydride at lysine residues. The intermolecular resonance energy transfer from tryptophan residues of
Sanjaya C Abeysirigunawardena et al.
Nature communications, 8(1), 492-492 (2017-09-10)
Assembly of 30S ribosomes involves the hierarchical addition of ribosomal proteins that progressively stabilize the folded 16S rRNA. Here, we use three-color single molecule FRET to show how combinations of ribosomal proteins uS4, uS17 and bS20 in the 16S 5'
Fariha Kanwal et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 48(5), 1915-1927 (2018-08-10)
RNA elements such as catalytic RNA, riboswitch, microRNA, and long non coding RNA (lncRNA) play central roles in many cellular processes. Studying diverse RNA functions require large quantities of RNA for precise structure analysis. Current RNA structure and function studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service