Skip to Content
Merck
All Photos(3)

Documents

127582

Sigma-Aldrich

N-(2-Hydroxyethyl)ethylenediamine

99%

Synonym(s):

2-(2-Aminoethylamino)ethanol, N-(2-Aminoethyl)ethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2NHCH2CH2OH
CAS Number:
Molecular Weight:
104.15
Beilstein:
506012
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.6 (vs air)

Quality Level

Assay

99%

refractive index

n20/D 1.485 (lit.)

bp

238-240 °C/752 mmHg (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

NCCNCCO

InChI

1S/C4H12N2O/c5-1-2-6-3-4-7/h6-7H,1-5H2

InChI key

LHIJANUOQQMGNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-(2-hydroxyethyl)ethylenediamine (hydeten) or [N-(2-aminoethyl)ethanolamine] can be used as:
  • A precursor in the synthesis of room-temperature ionic liquids, acetate and formate ammonium salts of N-(2-hydroxyethyl)-ethylenediamine.
  • A ligand in the preparation of polymeric cyano-bridged platinum(II)complexes, ligand-copper(II)carbohydrate complexes and cyano-bridged Ni-Pt and Cu-Pt coordination polymers.

It can also be used to study the aerobic biodecomposition of amines in hypersaline wastewaters.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

291.2 °F

Flash Point(C)

144 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Foti et al.
Contact dermatitis, 45(3), 129-133 (2001-09-13)
Amphoacetates are organic compounds used in many industrial applications and in cosmetic formulations for the skin, hair and mucosa, as surfactants, mild foaming and cleansing agents in concentrations ranging from 0.1% to 50%. Despite the fact that they have been
Steffen Schneider et al.
Birth defects research. Part B, Developmental and reproductive toxicology, 95(2), 95-106 (2012-03-31)
The reproductive and developmental toxicity of aminoethylethanolamine was evaluated in a standard screening study (OECD, 1995: Organisation for economic co-operation and development. Paris, France), in which groups of Wistar rats (10/sex/group) were administered the test substance by gavage at dosage
Investigation of Sugar-Binding Sites in Ternary Ligand- Copper (II)- Carbohydrate Complexes
Striegler S and Tewes E
European Journal of Inorganic Chemistry, 2002(2), 487-495 (2002)
Preparation, structural, magnetic and thermal properties of two heterobimetallic cyano-bridged nickel (II)-copper (II)/platinum (II) coordination polymers
Karadaug A, et al.
Inorgorganica Chimica Acta, 362(7), 2299-2304 (2009)
Ai-Ju Zhou et al.
Inorganic chemistry, 46(20), 8111-8113 (2007-09-11)
Room-temperature reactions of N,N,N',N'-tetrakis(2-hydroxyethyl)ethylenediamine with manganese(II) salts yield a novel family of Mn(12) clusters incorporating the same Mn(12)O(4) core and tunable oxidation states of Mn(III)(x)Mn(II)(12-x) (x = 8, 10, and 12). Magnetic susceptibility data indicate that the spin of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service