HPLC Analysis of Withania Extract on Ascentis® Express F5
Materials
analytical column
CONDITIONS
column
Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 μm particles (53569-U)
mobile phase
[A] water; [B] acetonitrile
gradient
20 to 100% B in 5 min; held at 100% B for 0.5 min
flow rate
0.3 mL/min
pressure
2750 psi (190 bar)
column temp.
35 °C
detector
UV, 227 nm
injection
5 μL
sample
20 mg/mL in water:acetonitrile (50:50); sonicate for 15 minutes; filter 0.45 μm; dilute to water:acetonitrile (80:20)
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express F5 for the efficient separation of withania extract.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany