Merck
  • A validated UHPLC method for the determination of caffeoylquinic and di-caffeoylquinic acids in green coffee extracts using an RP-Amide fused-core column.
A validated UHPLC method for the determination of caffeoylquinic and di-caffeoylquinic acids in green coffee extracts using an RP-Amide fused-core column.
Journal of pharmaceutical and biomedical analysis (2018-02-08)
Jakub Fibigr, Michaela Majorová, Hana Kočová Vlčková, Petr Solich, Dalibor Šatínský
ABSTRACT
The presented work describes the development and validation of a rapid UHPLC-UV method using a fused core particle column with an RP-Amide stationary phase for the separation and quantitative analysis of caffeoylquinic and di-caffeoylquinic acids in green coffee extracts. Three caffeoylquinic acids (3-caffeoylquinic acid, 4-caffeoylquinic acid, and 5-caffeoylquinic acid) and two di-caffeoylquinic acids (1,3-di-caffeoylquinic acid, and 3,5-di-caffeoylquinic acid) were separated and analyzed in 8 min. That was possible due to the unique selectivity of the RP-Amide stationary phase for the analyzed acids. The retention behavior of all analytes under different compositions of the mobile phase on different columns was evaluated in this study. The optimal chromatographic separation was performed using an Ascentis Express RP-Amide (100 × 2.1 mm) fused-core column with a particle size of 2.7 μm at a temperature of 30 °C. For validation of the newly developed method, acetonitrile was used as mobile phase B and 5% formic acid, filtrated through a 0.22 μm filter, was used as mobile phase A. They were delivered at a flow rate of 0.9 mL min
Product Description
Supelco
Ascentis® RP-Amide HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Ascentis® RP-Amide HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® RP-Amide Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Ascentis® RP-Amide HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Ascentis® RP-Amide HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Ascentis® RP-Amide Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
Supelco
Ascentis® RP-Amide HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® RP-Amide HPLC Column, 5 μm particle size, L × I.D. 15 cm × 21.2 mm
Supelco
Ascentis® RP-Amide Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm, pkg of 2 ea
Supelco
Ascentis® RP-Amide Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Ascentis® RP-Amide HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® RP-Amide HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Ascentis® RP-Amide HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 3 cm × 3 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 10 cm × 3 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 5 cm × 3 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Ascentis® Express RP-Amide, 5 μm Guard Cartridge, 5 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 3 cm × 4.6 mm
Supelco
Ascentis® RP-Amide HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
Ascentis® Express RP-Amide, 5 μm Guard Cartridge, 5 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea