Merck
  • [Determination of 32 sulfonylurea herbicide residues in sweet corns and green soybeans by QuEChERS-liquid chromatography-tandem mass spectrometry].
[Determination of 32 sulfonylurea herbicide residues in sweet corns and green soybeans by QuEChERS-liquid chromatography-tandem mass spectrometry].
Se pu = Chinese journal of chromatography (2015-09-22)
Lianzhu Wang, Xiaoyan Huang, Dengfei Wang, Yong Chen, Dunming Xu, Yu Zhou
ABSTRACT
A fast method based on QuEChERS methodology for the simultaneous determination of 32 sulfonylurea herbicide residues in sweet corns and green soybeans was developed using liquid chromatography-tandem mass spectrometry (LC-MS/MS). The clean-up effects of three dispersive sorbents were evaluated in terms of the residue mass for extracts after evaporation and recoveries. The three sorbents were C18, a mixture of two sorbents--silica coated with zirconium dioxide (Z-Sep) and C18, a bonded C18 zirconia-coated silica (Z-Sep+). As a result, the best effects were obtained from using Z-Sep/C18 sorbents. The samples were extracted with acetonitrile, and salted out with anhydrous magnesium sulphate and sodium chloride. The extracts were cleaned up with dispersive solid phase extraction using Z-Sep/C18 sorbents. Chromatographic analysis was carried out using a CSH C18 column with gradient elution. The pesticides were analyzed by negative electrospray ionization tandem mass spectrometry under scheduled multiple reaction monitoring mode. The quantification was achieved using matrix-matched standard calibrations as the external standard. The recoveries at fortification levels of 10, 20, 100 µg/kg in sweet corns and green beans ranged from 80.0% to 108.2% with the relative standard deviations of 1.2%-13.0%. The limits of quantification (S/N ≥ 10) were 0.2-5.0 µg/kg. The method has been proven to be simple, sensitive, environmental, and thus suitable for the determination of the 32 sulfonylurea herbicide residues in sweet corns and green soy- beans.
Product Description
Supelco
Supel QuE, Z-Sep/C18 Tube, centrifuge tube volume 2 mL, pack of 100
Supelco
Supel QuE, Z-Sep Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep+ Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep+ Bulk, pkg of 20 g
Supelco
Supel QuE, Z-Sep Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep+ Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep/C18 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep+ with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep Bulk